Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108634704-108634961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZIP3
Entrez Gene ID
9666
Gene Name
DAZ interacting zinc finger protein 3
Gene Aliases
PPP1R66, UURF2, hRUL138
UniGene
Hs.409210
Gene Chromosome Location
Chr.3: 108589490-108694846 on Build GRCh38
Transcripts
NM_014648.3 XM_005247914.3 XM_005247915.3 XM_005247916.3 XM_005247917.3 XM_005247918.3 XM_005247919.3 XM_017007540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5344292 c.837C>A p.C279* Chr.3: 108634891-108634891 on Build GRCh38
1536029 c.872G>T p.C291F Chr.3: 108634926-108634926 on Build GRCh38
1417645 c.877delA p.K294fs*5 Chr.3: 108634931-108634931 on Build GRCh38
2150885 c.818G>T p.G273V Chr.3: 108634872-108634872 on Build GRCh38
3426859 c.847T>C p.C283R Chr.3: 108634901-108634901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053571023 Chr.3: 108634893-108634893 on Build GRCh38
rs889742708 Chr.3: 108634775-108634775 on Build GRCh38
rs756329512 Chr.3: 108634836-108634836 on Build GRCh38
rs1034930283 Chr.3: 108634882-108634882 on Build GRCh38
rs1007217277 Chr.3: 108634776-108634776 on Build GRCh38
rs901358284 Chr.3: 108634873-108634874 on Build GRCh38
rs754868206 Chr.3: 108634820-108634820 on Build GRCh38
rs758290518 Chr.3: 108634826-108634826 on Build GRCh38
rs754208586 Chr.3: 108634771-108634771 on Build GRCh38
rs201059924 Chr.3: 108634888-108634888 on Build GRCh38
rs765056399 Chr.3: 108634822-108634822 on Build GRCh38
rs748627576 Chr.3: 108634856-108634856 on Build GRCh38
rs770684021 Chr.3: 108634925-108634925 on Build GRCh38
rs755299389 Chr.3: 108634790-108634790 on Build GRCh38
rs749634317 Chr.3: 108634843-108634843 on Build GRCh38
rs760319465 Chr.3: 108634903-108634903 on Build GRCh38
rs917468108 Chr.3: 108634912-108634912 on Build GRCh38
rs781276207 Chr.3: 108634834-108634834 on Build GRCh38
rs200630414 Chr.3: 108634899-108634899 on Build GRCh38
rs750510754 Chr.3: 108634835-108634835 on Build GRCh38
rs776464139 Chr.3: 108634927-108634927 on Build GRCh38
rs768965325 Chr.3: 108634853-108634853 on Build GRCh38
rs765759570 Chr.3: 108634794-108634794 on Build GRCh38
rs774054237 Chr.3: 108634859-108634859 on Build GRCh38
rs993811027 Chr.3: 108634813-108634813 on Build GRCh38
rs780315369 Chr.3: 108634838-108634838 on Build GRCh38
rs752643043 Chr.3: 108634824-108634824 on Build GRCh38
rs1048132358 Chr.3: 108634802-108634802 on Build GRCh38
rs758910567 Chr.3: 108634860-108634860 on Build GRCh38

View Full Product Details