Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37574232-37574483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL2
Entrez Gene ID
29889
Gene Name
G protein nucleolar 2
Gene Aliases
HUMAUANTIG, NGP1, Ngp-1, Nog2, Nug2
UniGene
Hs.75528
Gene Chromosome Location
Chr.1: 37566812-37595985 on Build GRCh38
Transcripts
NM_001323623.1 NM_001323624.1 NM_013285.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1687476 c.1315G>A p.D439N Chr.1: 37574444-37574444 on Build GRCh38
3489211 c.1370C>T p.P457L Chr.1: 37574389-37574389 on Build GRCh38
7164076 c.1414C>G p.Q472E Chr.1: 37574345-37574345 on Build GRCh38
426212 c.1359G>T p.R453S Chr.1: 37574400-37574400 on Build GRCh38
6445468 c.1374C>G p.F458L Chr.1: 37574385-37574385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369456784 Chr.1: 37574415-37574415 on Build GRCh38
rs754553486 Chr.1: 37574412-37574412 on Build GRCh38
rs749107201 Chr.1: 37574347-37574347 on Build GRCh38
rs760784694 Chr.1: 37574371-37574371 on Build GRCh38
rs747150226 Chr.1: 37574457-37574458 on Build GRCh38
rs758051388 Chr.1: 37574303-37574303 on Build GRCh38
rs775671790 Chr.1: 37574365-37574365 on Build GRCh38
rs758338395 Chr.1: 37574344-37574345 on Build GRCh38
rs148607956 Chr.1: 37574396-37574396 on Build GRCh38
rs546299461 Chr.1: 37574326-37574326 on Build GRCh38
rs148065418 Chr.1: 37574265-37574265 on Build GRCh38
rs188720818 Chr.1: 37574257-37574257 on Build GRCh38
rs768683012 Chr.1: 37574385-37574385 on Build GRCh38
rs905574931 Chr.1: 37574351-37574351 on Build GRCh38
rs200697753 Chr.1: 37574310-37574310 on Build GRCh38
rs756075370 Chr.1: 37574420-37574420 on Build GRCh38
rs777147669 Chr.1: 37574389-37574389 on Build GRCh38
rs772244867 Chr.1: 37574356-37574356 on Build GRCh38
rs754506258 Chr.1: 37574313-37574313 on Build GRCh38
rs375305276 Chr.1: 37574445-37574445 on Build GRCh38
rs781305406 Chr.1: 37574320-37574320 on Build GRCh38
rs752683186 Chr.1: 37574418-37574418 on Build GRCh38
rs777367193 Chr.1: 37574328-37574328 on Build GRCh38
rs12025870 Chr.1: 37574403-37574403 on Build GRCh38
rs1034019898 Chr.1: 37574270-37574270 on Build GRCh38
rs765472280 Chr.1: 37574297-37574297 on Build GRCh38
rs750809725 Chr.1: 37574395-37574395 on Build GRCh38
rs866270321 Chr.1: 37574382-37574382 on Build GRCh38
rs751213477 Chr.1: 37574405-37574405 on Build GRCh38
rs75520059 Chr.1: 37574280-37574280 on Build GRCh38
rs984284001 Chr.1: 37574264-37574264 on Build GRCh38
rs748044918 Chr.1: 37574324-37574324 on Build GRCh38
rs992424553 Chr.1: 37574294-37574294 on Build GRCh38
rs757671599 Chr.1: 37574295-37574295 on Build GRCh38
rs750105572 Chr.1: 37574299-37574299 on Build GRCh38
rs780053113 Chr.1: 37574345-37574345 on Build GRCh38
rs991000048 Chr.1: 37574287-37574287 on Build GRCh38
rs751164174 Chr.1: 37574312-37574312 on Build GRCh38
rs374936102 Chr.1: 37574258-37574258 on Build GRCh38
rs12025869 Chr.1: 37574401-37574401 on Build GRCh38
rs373181030 Chr.1: 37574444-37574444 on Build GRCh38

View Full Product Details