Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155317909-155318170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDPS
Entrez Gene ID
2224
Gene Name
farnesyl diphosphate synthase
Gene Aliases
FPPS, FPS, POROK9
UniGene
Hs.335918
Gene Chromosome Location
Chr.1: 155308748-155320666 on Build GRCh38
Transcripts
NM_001135821.1 NM_001135822.1 NM_001242824.1 NM_001242825.1 NM_002004.3 XM_005244962.1 XM_005244963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6662994 c.563C>T p.P188L Chr.1: 155318170-155318170 on Build GRCh38
4135573 c.486A>G p.Q162Q Chr.1: 155317946-155317946 on Build GRCh38
2153567 c.532C>T p.R178C Chr.1: 155317992-155317992 on Build GRCh38
896880 c.537G>T p.R179R Chr.1: 155317997-155317997 on Build GRCh38
4833994 c.556C>G p.Q186E Chr.1: 155318016-155318016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530418099 Chr.1: 155318121-155318121 on Build GRCh38
rs1020784588 Chr.1: 155318080-155318080 on Build GRCh38
rs749581999 Chr.1: 155317992-155317992 on Build GRCh38
rs966601778 Chr.1: 155318101-155318101 on Build GRCh38
rs372604047 Chr.1: 155318071-155318071 on Build GRCh38
rs746349237 Chr.1: 155318006-155318006 on Build GRCh38
rs745999713 Chr.1: 155318052-155318053 on Build GRCh38
rs374801518 Chr.1: 155318026-155318026 on Build GRCh38
rs761596390 Chr.1: 155318073-155318073 on Build GRCh38
rs199833424 Chr.1: 155318126-155318126 on Build GRCh38
rs999731346 Chr.1: 155318103-155318103 on Build GRCh38
rs771194005 Chr.1: 155317993-155317993 on Build GRCh38
rs772516967 Chr.1: 155318011-155318011 on Build GRCh38
rs758451788 Chr.1: 155317946-155317946 on Build GRCh38
rs772800704 Chr.1: 155318134-155318134 on Build GRCh38
rs962750064 Chr.1: 155317937-155317937 on Build GRCh38
rs863225241 Chr.1: 155317996-155317996 on Build GRCh38
rs777799727 Chr.1: 155317990-155317990 on Build GRCh38
rs908635864 Chr.1: 155317936-155317936 on Build GRCh38
rs768229747 Chr.1: 155318052-155318052 on Build GRCh38
rs114439752 Chr.1: 155318045-155318045 on Build GRCh38
rs533042404 Chr.1: 155317977-155317977 on Build GRCh38
rs751554976 Chr.1: 155318141-155318141 on Build GRCh38
rs1050358148 Chr.1: 155318035-155318035 on Build GRCh38
rs752776897 Chr.1: 155317981-155317981 on Build GRCh38
rs879528394 Chr.1: 155318061-155318061 on Build GRCh38
rs189381460 Chr.1: 155318109-155318109 on Build GRCh38
rs932949076 Chr.1: 155317999-155317999 on Build GRCh38
rs141830995 Chr.1: 155318125-155318125 on Build GRCh38
rs756295209 Chr.1: 155317985-155317985 on Build GRCh38
rs1011640540 Chr.1: 155318068-155318068 on Build GRCh38
rs762376502 Chr.1: 155317998-155317998 on Build GRCh38

View Full Product Details