Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50378775-50379028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EME1
Entrez Gene ID
146956
Gene Name
essential meiotic structure-specific endonuclease 1
Gene Aliases
MMS4L, SLX2A
UniGene
Hs.514330
Gene Chromosome Location
Chr.17: 50373220-50381479 on Build GRCh38
Transcripts
NM_001166131.1 NM_152463.2 XM_005257081.3 XM_011524395.2 XM_017024236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
148297 c.1049T>C p.I350T Chr.17: 50378832-50378832 on Build GRCh38
7276037 c.1079C>T p.S360L Chr.17: 50378862-50378862 on Build GRCh38
3362438 c.1103A>T p.K368I Chr.17: 50378886-50378886 on Build GRCh38
981007 c.1073_1074delCT p.L359fs*26 Chr.17: 50378856-50378857 on Build GRCh38
5767262 c.1075C>G p.L359V Chr.17: 50378858-50378858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749928401 Chr.17: 50378845-50378845 on Build GRCh38
rs371452958 Chr.17: 50378869-50378869 on Build GRCh38
rs756338548 Chr.17: 50378839-50378839 on Build GRCh38
rs76292406 Chr.17: 50378938-50378938 on Build GRCh38
rs778593353 Chr.17: 50378814-50378815 on Build GRCh38
rs778157276 Chr.17: 50378855-50378855 on Build GRCh38
rs7222520 Chr.17: 50378822-50378822 on Build GRCh38
rs746436410 Chr.17: 50378891-50378891 on Build GRCh38
rs12450550 Chr.17: 50378832-50378832 on Build GRCh38
rs199942900 Chr.17: 50378844-50378844 on Build GRCh38
rs993468388 Chr.17: 50378953-50378953 on Build GRCh38
rs998313887 Chr.17: 50378997-50378997 on Build GRCh38
rs148232687 Chr.17: 50378884-50378884 on Build GRCh38
rs752963366 Chr.17: 50378838-50378838 on Build GRCh38
rs888053295 Chr.17: 50378984-50378984 on Build GRCh38
rs376696640 Chr.17: 50378913-50378913 on Build GRCh38
rs761507904 Chr.17: 50378803-50378803 on Build GRCh38
rs938487188 Chr.17: 50378856-50378856 on Build GRCh38
rs769569148 Chr.17: 50378804-50378804 on Build GRCh38
rs772921012 Chr.17: 50378808-50378808 on Build GRCh38
rs1039731626 Chr.17: 50378952-50378952 on Build GRCh38
rs73346027 Chr.17: 50378932-50378932 on Build GRCh38
rs780744447 Chr.17: 50378903-50378903 on Build GRCh38
rs756880780 Chr.17: 50378812-50378813 on Build GRCh38
rs767764539 Chr.17: 50378837-50378837 on Build GRCh38
rs143055740 Chr.17: 50378835-50378835 on Build GRCh38
rs535766431 Chr.17: 50378809-50378809 on Build GRCh38
rs769376064 Chr.17: 50378910-50378910 on Build GRCh38
rs17714940 Chr.17: 50378890-50378890 on Build GRCh38
rs901165779 Chr.17: 50378957-50378957 on Build GRCh38
rs186599955 Chr.17: 50378886-50378886 on Build GRCh38
rs565863242 Chr.17: 50379007-50379007 on Build GRCh38
rs913411946 Chr.17: 50378900-50378900 on Build GRCh38
rs757909327 Chr.17: 50378947-50378948 on Build GRCh38
rs201617846 Chr.17: 50378893-50378893 on Build GRCh38
rs374070235 Chr.17: 50378904-50378904 on Build GRCh38
rs749008953 Chr.17: 50378931-50378931 on Build GRCh38
rs947285448 Chr.17: 50378888-50378888 on Build GRCh38

View Full Product Details