Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36244480-36244753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAR2
Entrez Gene ID
25980
Gene Name
AAR2 splicing factor homolog
Gene Aliases
C20orf4, CGI-23
UniGene
Hs.11314
Gene Chromosome Location
Chr.20: 36236136-36256941 on Build GRCh38
Transcripts
NM_001271874.1 NM_015511.4 XM_006723770.3 XM_011528762.2 XM_011528763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5744042 c.774delT p.F259fs*20 Chr.20: 36244713-36244713 on Build GRCh38
359685 c.801G>T p.V267V Chr.20: 36244740-36244740 on Build GRCh38
5731199 c.763C>T p.L255F Chr.20: 36244702-36244702 on Build GRCh38
5631179 c.805G>A p.E269K Chr.20: 36244744-36244744 on Build GRCh38
5974016 c.804C>T p.Y268Y Chr.20: 36244743-36244743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006565148 Chr.20: 36244552-36244552 on Build GRCh38
rs965067879 Chr.20: 36244592-36244592 on Build GRCh38
rs776514716 Chr.20: 36244677-36244679 on Build GRCh38
rs753691544 Chr.20: 36244705-36244705 on Build GRCh38
rs532751767 Chr.20: 36244537-36244537 on Build GRCh38
rs543945528 Chr.20: 36244513-36244513 on Build GRCh38
rs918316408 Chr.20: 36244608-36244608 on Build GRCh38
rs760425982 Chr.20: 36244702-36244702 on Build GRCh38
rs112647820 Chr.20: 36244681-36244682 on Build GRCh38
rs368599383 Chr.20: 36244716-36244716 on Build GRCh38
rs767054441 Chr.20: 36244683-36244683 on Build GRCh38
rs770205897 Chr.20: 36244704-36244704 on Build GRCh38
rs928542035 Chr.20: 36244602-36244602 on Build GRCh38
rs980014886 Chr.20: 36244598-36244598 on Build GRCh38
rs562607102 Chr.20: 36244528-36244528 on Build GRCh38
rs771074176 Chr.20: 36244676-36244680 on Build GRCh38
rs1032274733 Chr.20: 36244518-36244518 on Build GRCh38
rs1043152983 Chr.20: 36244684-36244684 on Build GRCh38
rs35283785 Chr.20: 36244680-36244681 on Build GRCh38
rs953004216 Chr.20: 36244625-36244625 on Build GRCh38
rs763925130 Chr.20: 36244704-36244704 on Build GRCh38
rs759141865 Chr.20: 36244673-36244673 on Build GRCh38
rs1000680916 Chr.20: 36244509-36244509 on Build GRCh38
rs141514980 Chr.20: 36244646-36244646 on Build GRCh38
rs115016138 Chr.20: 36244579-36244579 on Build GRCh38
rs547595252 Chr.20: 36244526-36244526 on Build GRCh38
rs568801996 Chr.20: 36244653-36244655 on Build GRCh38
rs867724451 Chr.20: 36244705-36244705 on Build GRCh38
rs1057307448 Chr.20: 36244711-36244711 on Build GRCh38
rs984269726 Chr.20: 36244641-36244641 on Build GRCh38
rs397864454 Chr.20: 36244684-36244685 on Build GRCh38
rs752437824 Chr.20: 36244690-36244690 on Build GRCh38
rs972925602 Chr.20: 36244596-36244596 on Build GRCh38

View Full Product Details