Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46360475-46360744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPAS1
Entrez Gene ID
2034
Gene Name
endothelial PAS domain protein 1
Gene Aliases
ECYT4, HIF2A, HLF, MOP2, PASD2, bHLHe73
UniGene
Hs.468410
Gene Chromosome Location
Chr.2: 46297402-46386703 on Build GRCh38
Transcripts
NM_001430.4 XM_011532698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5049219 c.482A>G p.D161G Chr.2: 46360665-46360665 on Build GRCh38
477449 c.497G>T p.R166L Chr.2: 46360680-46360680 on Build GRCh38
6654531 c.496C>T p.R166W Chr.2: 46360679-46360679 on Build GRCh38
4681151 c.469delA p.S159fs*12 Chr.2: 46360652-46360652 on Build GRCh38
442951 c.493G>C p.E165Q Chr.2: 46360676-46360676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190619488 Chr.2: 46360579-46360579 on Build GRCh38
rs115264325 Chr.2: 46360708-46360708 on Build GRCh38
rs141965374 Chr.2: 46360664-46360664 on Build GRCh38
rs141537861 Chr.2: 46360629-46360629 on Build GRCh38
rs760079550 Chr.2: 46360602-46360602 on Build GRCh38
rs116249013 Chr.2: 46360616-46360616 on Build GRCh38
rs759884897 Chr.2: 46360706-46360706 on Build GRCh38
rs765172544 Chr.2: 46360656-46360664 on Build GRCh38
rs186236229 Chr.2: 46360505-46360505 on Build GRCh38
rs778655161 Chr.2: 46360660-46360660 on Build GRCh38
rs76884025 Chr.2: 46360667-46360667 on Build GRCh38
rs1027571167 Chr.2: 46360690-46360690 on Build GRCh38
rs755022508 Chr.2: 46360658-46360658 on Build GRCh38
rs747284735 Chr.2: 46360668-46360668 on Build GRCh38
rs761418476 Chr.2: 46360717-46360717 on Build GRCh38
rs371949833 Chr.2: 46360589-46360589 on Build GRCh38
rs1023662517 Chr.2: 46360552-46360552 on Build GRCh38
rs751141124 Chr.2: 46360609-46360609 on Build GRCh38
rs755805398 Chr.2: 46360634-46360634 on Build GRCh38
rs889262086 Chr.2: 46360543-46360543 on Build GRCh38
rs772001481 Chr.2: 46360666-46360666 on Build GRCh38
rs182405852 Chr.2: 46360584-46360584 on Build GRCh38
rs777049674 Chr.2: 46360677-46360677 on Build GRCh38
rs1050674628 Chr.2: 46360524-46360524 on Build GRCh38
rs539767978 Chr.2: 46360559-46360559 on Build GRCh38
rs753679311 Chr.2: 46360657-46360657 on Build GRCh38
rs765660809 Chr.2: 46360603-46360603 on Build GRCh38
rs374874715 Chr.2: 46360592-46360592 on Build GRCh38
rs766889111 Chr.2: 46360721-46360721 on Build GRCh38
rs576804215 Chr.2: 46360509-46360509 on Build GRCh38
rs771115759 Chr.2: 46360670-46360670 on Build GRCh38
rs750004234 Chr.2: 46360631-46360631 on Build GRCh38
rs1013166026 Chr.2: 46360529-46360529 on Build GRCh38
rs377257704 Chr.2: 46360649-46360649 on Build GRCh38
rs576196431 Chr.2: 46360583-46360583 on Build GRCh38

View Full Product Details