Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38938518-38938791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO47
Entrez Gene ID
494188
Gene Name
F-box protein 47
Gene Aliases
-
UniGene
Hs.549536
Gene Chromosome Location
Chr.17: 38936278-38967475 on Build GRCh38
Transcripts
NM_001008777.2 XM_011524865.2 XM_011524866.2 XM_011524867.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4594951 c.1210G>T p.V404F Chr.17: 38938606-38938606 on Build GRCh38
161019 c.1128G>A p.M376I Chr.17: 38938688-38938688 on Build GRCh38
6146711 c.1147G>C p.V383L Chr.17: 38938669-38938669 on Build GRCh38
4851991 c.1090G>C p.E364Q Chr.17: 38938726-38938726 on Build GRCh38
3516734 c.1220A>T p.E407V Chr.17: 38938596-38938596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565388199 Chr.17: 38938577-38938577 on Build GRCh38
rs1055235563 Chr.17: 38938542-38938542 on Build GRCh38
rs550325023 Chr.17: 38938547-38938547 on Build GRCh38
rs757390672 Chr.17: 38938551-38938551 on Build GRCh38
rs752719743 Chr.17: 38938752-38938752 on Build GRCh38
rs770298267 Chr.17: 38938619-38938619 on Build GRCh38
rs758904291 Chr.17: 38938611-38938611 on Build GRCh38
rs181650182 Chr.17: 38938545-38938545 on Build GRCh38
rs754326979 Chr.17: 38938675-38938675 on Build GRCh38
rs781385307 Chr.17: 38938556-38938556 on Build GRCh38
rs776234579 Chr.17: 38938608-38938608 on Build GRCh38
rs750523882 Chr.17: 38938711-38938711 on Build GRCh38
rs974461292 Chr.17: 38938651-38938651 on Build GRCh38
rs748081611 Chr.17: 38938579-38938579 on Build GRCh38
rs1008403490 Chr.17: 38938601-38938601 on Build GRCh38
rs780272947 Chr.17: 38938747-38938747 on Build GRCh38
rs187260473 Chr.17: 38938664-38938664 on Build GRCh38
rs746720913 Chr.17: 38938589-38938589 on Build GRCh38
rs767597968 Chr.17: 38938708-38938708 on Build GRCh38
rs527875930 Chr.17: 38938544-38938544 on Build GRCh38
rs370299590 Chr.17: 38938540-38938540 on Build GRCh38
rs148455055 Chr.17: 38938704-38938704 on Build GRCh38
rs756202363 Chr.17: 38938730-38938730 on Build GRCh38
rs371873841 Chr.17: 38938753-38938753 on Build GRCh38
rs776035859 Chr.17: 38938646-38938646 on Build GRCh38
rs532833037 Chr.17: 38938582-38938582 on Build GRCh38
rs746536363 Chr.17: 38938594-38938594 on Build GRCh38
rs373996927 Chr.17: 38938606-38938606 on Build GRCh38
rs745888562 Chr.17: 38938567-38938567 on Build GRCh38
rs145585502 Chr.17: 38938685-38938685 on Build GRCh38
rs754978265 Chr.17: 38938575-38938575 on Build GRCh38
rs777678086 Chr.17: 38938583-38938583 on Build GRCh38

View Full Product Details