Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21178160-21178415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7B
Entrez Gene ID
25979
Gene Name
dehydrogenase/reductase 7B
Gene Aliases
CGI-93, SDR32C1
UniGene
Hs.386989
Gene Chromosome Location
Chr.17: 21123369-21191972 on Build GRCh38
Transcripts
NM_015510.4 XM_005256595.4 XM_005256596.3 XM_011523786.2 XM_017024425.1 XM_017024426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
976571 c.283G>T p.E95* Chr.17: 21178316-21178316 on Build GRCh38
3515132 c.271G>A p.E91K Chr.17: 21178304-21178304 on Build GRCh38
4410507 c.225G>A p.A75A Chr.17: 21178258-21178258 on Build GRCh38
976570 c.260G>T p.G87V Chr.17: 21178293-21178293 on Build GRCh38
976572 c.292G>A p.A98T Chr.17: 21178325-21178325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371131037 Chr.17: 21178382-21178382 on Build GRCh38
rs750247244 Chr.17: 21178325-21178325 on Build GRCh38
rs779894265 Chr.17: 21178191-21178191 on Build GRCh38
rs538890481 Chr.17: 21178214-21178215 on Build GRCh38
rs755919181 Chr.17: 21178342-21178342 on Build GRCh38
rs370358348 Chr.17: 21178333-21178333 on Build GRCh38
rs779665675 Chr.17: 21178296-21178296 on Build GRCh38
rs920853495 Chr.17: 21178301-21178301 on Build GRCh38
rs539133971 Chr.17: 21178186-21178186 on Build GRCh38
rs768598719 Chr.17: 21178216-21178216 on Build GRCh38
rs757076694 Chr.17: 21178257-21178257 on Build GRCh38
rs1017736530 Chr.17: 21178331-21178331 on Build GRCh38
rs199649455 Chr.17: 21178283-21178283 on Build GRCh38
rs765598028 Chr.17: 21178273-21178273 on Build GRCh38
rs116227155 Chr.17: 21178323-21178323 on Build GRCh38
rs199845654 Chr.17: 21178258-21178258 on Build GRCh38
rs748124837 Chr.17: 21178245-21178245 on Build GRCh38
rs868187094 Chr.17: 21178281-21178281 on Build GRCh38
rs116321095 Chr.17: 21178393-21178393 on Build GRCh38
rs780756368 Chr.17: 21178183-21178183 on Build GRCh38
rs143021440 Chr.17: 21178356-21178356 on Build GRCh38
rs771972829 Chr.17: 21178246-21178246 on Build GRCh38
rs370002673 Chr.17: 21178297-21178297 on Build GRCh38
rs776634023 Chr.17: 21178379-21178379 on Build GRCh38
rs559570913 Chr.17: 21178392-21178392 on Build GRCh38
rs775760613 Chr.17: 21178384-21178384 on Build GRCh38
rs115536974 Chr.17: 21178286-21178286 on Build GRCh38
rs780031124 Chr.17: 21178350-21178350 on Build GRCh38
rs190857040 Chr.17: 21178235-21178235 on Build GRCh38
rs368561645 Chr.17: 21178385-21178385 on Build GRCh38
rs752075245 Chr.17: 21178290-21178290 on Build GRCh38
rs764479723 Chr.17: 21178391-21178391 on Build GRCh38
rs116883245 Chr.17: 21178366-21178366 on Build GRCh38
rs773060534 Chr.17: 21178355-21178355 on Build GRCh38
rs755717279 Chr.17: 21178291-21178291 on Build GRCh38
rs373237004 Chr.17: 21178213-21178213 on Build GRCh38
rs558138265 Chr.17: 21178296-21178296 on Build GRCh38
rs377203212 Chr.17: 21178251-21178251 on Build GRCh38
rs755664490 Chr.17: 21178188-21178188 on Build GRCh38
rs748909477 Chr.17: 21178192-21178192 on Build GRCh38
rs781185994 Chr.17: 21178260-21178260 on Build GRCh38
rs142632890 Chr.17: 21178300-21178300 on Build GRCh38
rs758778438 Chr.17: 21178284-21178284 on Build GRCh38
rs746111711 Chr.17: 21178264-21178265 on Build GRCh38

View Full Product Details