Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134036035-134036290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD3
Entrez Gene ID
8019
Gene Name
bromodomain containing 3
Gene Aliases
ORFX, RING3L
UniGene
Hs.522472
Gene Chromosome Location
Chr.9: 134030305-134068533 on Build GRCh38
Transcripts
NM_007371.3 XM_006717291.2 XM_011519052.2 XM_017015165.1 XM_017015166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5766088 c.1880G>A p.R627Q Chr.9: 134036088-134036088 on Build GRCh38
6612642 c.1722C>T p.Y574Y Chr.9: 134036246-134036246 on Build GRCh38
6582192 c.1696G>C p.E566Q Chr.9: 134036272-134036272 on Build GRCh38
5635829 c.1833C>T p.P611P Chr.9: 134036135-134036135 on Build GRCh38
1674941 c.1826C>T p.S609F Chr.9: 134036142-134036142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896277363 Chr.9: 134036195-134036195 on Build GRCh38
rs758351422 Chr.9: 134036263-134036265 on Build GRCh38
rs201071148 Chr.9: 134036134-134036134 on Build GRCh38
rs1002210873 Chr.9: 134036250-134036250 on Build GRCh38
rs200686859 Chr.9: 134036204-134036204 on Build GRCh38
rs777297159 Chr.9: 134036185-134036185 on Build GRCh38
rs201595589 Chr.9: 134036153-134036153 on Build GRCh38
rs757843939 Chr.9: 134036168-134036168 on Build GRCh38
rs763276141 Chr.9: 134036261-134036261 on Build GRCh38
rs754769412 Chr.9: 134036154-134036154 on Build GRCh38
rs1045137667 Chr.9: 134036218-134036218 on Build GRCh38
rs201885398 Chr.9: 134036105-134036105 on Build GRCh38
rs367551230 Chr.9: 134036061-134036061 on Build GRCh38
rs952178417 Chr.9: 134036262-134036262 on Build GRCh38
rs778623028 Chr.9: 134036162-134036162 on Build GRCh38
rs766035056 Chr.9: 134036141-134036141 on Build GRCh38
rs909070256 Chr.9: 134036076-134036076 on Build GRCh38
rs550352346 Chr.9: 134036089-134036089 on Build GRCh38
rs200111572 Chr.9: 134036156-134036156 on Build GRCh38
rs55944890 Chr.9: 134036243-134036243 on Build GRCh38
rs139574233 Chr.9: 134036260-134036260 on Build GRCh38
rs941834319 Chr.9: 134036120-134036120 on Build GRCh38
rs201389658 Chr.9: 134036246-134036246 on Build GRCh38
rs371391133 Chr.9: 134036166-134036166 on Build GRCh38
rs201112315 Chr.9: 134036192-134036192 on Build GRCh38
rs768813177 Chr.9: 134036263-134036263 on Build GRCh38
rs202106159 Chr.9: 134036211-134036211 on Build GRCh38
rs369709520 Chr.9: 134036135-134036135 on Build GRCh38
rs761403724 Chr.9: 134036188-134036188 on Build GRCh38
rs749966068 Chr.9: 134036257-134036257 on Build GRCh38
rs448698 Chr.9: 134036132-134036132 on Build GRCh38
rs770116899 Chr.9: 134036258-134036258 on Build GRCh38

View Full Product Details