Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103382433-103382664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTHRC1
Entrez Gene ID
115908
Gene Name
collagen triple helix repeat containing 1
Gene Aliases
-
UniGene
Hs.405614
Gene Chromosome Location
Chr.8: 103371515-103383005 on Build GRCh38
Transcripts
NM_001256099.1 NM_138455.3 XM_011516824.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7427822 c.681T>A p.S227S Chr.8: 103382549-103382549 on Build GRCh38
5515271 c.689A>C p.*230S Chr.8: 103382599-103382599 on Build GRCh38
3643394 c.582A>G p.L194L Chr.8: 103382492-103382492 on Build GRCh38
6849066 c.599T>G p.L200R Chr.8: 103382467-103382467 on Build GRCh38
6319734 c.550G>T p.E184* Chr.8: 103382460-103382460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984795003 Chr.8: 103382627-103382627 on Build GRCh38
rs74496760 Chr.8: 103382476-103382476 on Build GRCh38
rs754161630 Chr.8: 103382576-103382576 on Build GRCh38
rs778614680 Chr.8: 103382515-103382515 on Build GRCh38
rs767599276 Chr.8: 103382485-103382485 on Build GRCh38
rs200589758 Chr.8: 103382569-103382569 on Build GRCh38
rs762922336 Chr.8: 103382626-103382627 on Build GRCh38
rs756063602 Chr.8: 103382496-103382496 on Build GRCh38
rs1050134230 Chr.8: 103382487-103382487 on Build GRCh38
rs143595509 Chr.8: 103382550-103382550 on Build GRCh38
rs777739644 Chr.8: 103382502-103382502 on Build GRCh38
rs769689069 Chr.8: 103382607-103382608 on Build GRCh38
rs752656527 Chr.8: 103382488-103382488 on Build GRCh38
rs774596955 Chr.8: 103382587-103382587 on Build GRCh38
rs771540899 Chr.8: 103382524-103382524 on Build GRCh38
rs901985226 Chr.8: 103382477-103382477 on Build GRCh38
rs752783129 Chr.8: 103382634-103382634 on Build GRCh38
rs3098233 Chr.8: 103382516-103382516 on Build GRCh38
rs781701664 Chr.8: 103382528-103382528 on Build GRCh38
rs375851229 Chr.8: 103382578-103382578 on Build GRCh38
rs753600909 Chr.8: 103382507-103382507 on Build GRCh38
rs773573431 Chr.8: 103382565-103382565 on Build GRCh38
rs888869193 Chr.8: 103382498-103382498 on Build GRCh38
rs770249018 Chr.8: 103382552-103382552 on Build GRCh38
rs751608144 Chr.8: 103382479-103382479 on Build GRCh38
rs958816850 Chr.8: 103382622-103382622 on Build GRCh38
rs868734430 Chr.8: 103382580-103382580 on Build GRCh38
rs759580077 Chr.8: 103382613-103382613 on Build GRCh38
rs767759004 Chr.8: 103382572-103382572 on Build GRCh38
rs927379754 Chr.8: 103382611-103382611 on Build GRCh38
rs752715912 Chr.8: 103382563-103382563 on Build GRCh38
rs956632343 Chr.8: 103382518-103382518 on Build GRCh38
rs767725797 Chr.8: 103382623-103382623 on Build GRCh38
rs763322692 Chr.8: 103382571-103382571 on Build GRCh38
rs775288902 Chr.8: 103382611-103382612 on Build GRCh38
rs35877609 Chr.8: 103382482-103382482 on Build GRCh38
rs147418120 Chr.8: 103382514-103382514 on Build GRCh38
rs759391397 Chr.8: 103382494-103382494 on Build GRCh38
rs760692453 Chr.8: 103382639-103382639 on Build GRCh38
rs756872409 Chr.8: 103382508-103382508 on Build GRCh38

View Full Product Details