Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65669854-65670117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4A
Entrez Gene ID
10260
Gene Name
DENN domain containing 4A
Gene Aliases
IRLB, MYCPBP
UniGene
Hs.654567
Gene Chromosome Location
Chr.15: 65659123-65792293 on Build GRCh38
Transcripts
NM_001144823.2 NM_001320835.1 NM_005848.4 XM_005254120.4 XM_005254121.3 XM_005254122.4 XM_006720371.3 XM_011521157.2 XM_017021862.1 XM_017021863.1 XM_017021864.1 XM_017021865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4056341 c.4575C>T p.F1525F Chr.15: 65670075-65670075 on Build GRCh38
6642385 c.4435A>G p.T1479A Chr.15: 65670086-65670086 on Build GRCh38
5362985 c.4533C>T p.G1511G Chr.15: 65670117-65670117 on Build GRCh38
1134018 c.4609G>C p.E1537Q Chr.15: 65670041-65670041 on Build GRCh38
4056340 c.4628G>A p.R1543Q Chr.15: 65670022-65670022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761834858 Chr.15: 65669906-65669906 on Build GRCh38
rs760850251 Chr.15: 65669882-65669882 on Build GRCh38
rs762837882 Chr.15: 65670082-65670082 on Build GRCh38
rs750405918 Chr.15: 65670069-65670069 on Build GRCh38
rs376663396 Chr.15: 65669962-65669962 on Build GRCh38
rs746182065 Chr.15: 65669984-65669984 on Build GRCh38
rs760941399 Chr.15: 65670004-65670004 on Build GRCh38
rs902854583 Chr.15: 65670092-65670092 on Build GRCh38
rs750180940 Chr.15: 65669937-65669937 on Build GRCh38
rs775769252 Chr.15: 65669999-65669999 on Build GRCh38
rs1046467985 Chr.15: 65669935-65669935 on Build GRCh38
rs765266078 Chr.15: 65669923-65669923 on Build GRCh38
rs928074098 Chr.15: 65669943-65669943 on Build GRCh38
rs765214676 Chr.15: 65670044-65670044 on Build GRCh38
rs769064520 Chr.15: 65670009-65670009 on Build GRCh38
rs761994040 Chr.15: 65670023-65670023 on Build GRCh38
rs780665043 Chr.15: 65669951-65669951 on Build GRCh38
rs760034089 Chr.15: 65669995-65669995 on Build GRCh38
rs546711194 Chr.15: 65669978-65669978 on Build GRCh38
rs747663704 Chr.15: 65669976-65669976 on Build GRCh38
rs754026101 Chr.15: 65669904-65669904 on Build GRCh38
rs1055665654 Chr.15: 65669973-65669973 on Build GRCh38
rs764215682 Chr.15: 65669890-65669890 on Build GRCh38
rs965338340 Chr.15: 65670070-65670070 on Build GRCh38
rs772455780 Chr.15: 65669992-65669992 on Build GRCh38
rs758233842 Chr.15: 65669939-65669939 on Build GRCh38
rs754636173 Chr.15: 65669945-65669945 on Build GRCh38
rs369909860 Chr.15: 65670022-65670022 on Build GRCh38
rs367630037 Chr.15: 65670084-65670084 on Build GRCh38
rs1011517191 Chr.15: 65670067-65670067 on Build GRCh38
rs780677372 Chr.15: 65669964-65669964 on Build GRCh38
rs1019861154 Chr.15: 65670064-65670064 on Build GRCh38

View Full Product Details