Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48470416-48470651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH2
Entrez Gene ID
9266
Gene Name
cytohesin 2
Gene Aliases
ARNO, CTS18, CTS18.1, PSCD2, PSCD2L, SEC7L, Sec7p-L, Sec7p-like
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48469208-48482314 on Build GRCh38
Transcripts
NM_004228.6 NM_017457.5 XM_006723472.2 XM_006723473.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6860142 c.154G>A p.A52T Chr.19: 48470487-48470487 on Build GRCh38
6639561 c.109C>T p.R37W Chr.19: 48470442-48470442 on Build GRCh38
6639563 c.136G>A p.E46K Chr.19: 48470469-48470469 on Build GRCh38
4676060 c.128C>A p.A43D Chr.19: 48470461-48470461 on Build GRCh38
3107410 c.165C>T p.G55G Chr.19: 48470498-48470498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773173250 Chr.19: 48470542-48470542 on Build GRCh38
rs893860978 Chr.19: 48470479-48470479 on Build GRCh38
rs759883009 Chr.19: 48470487-48470487 on Build GRCh38
rs751989393 Chr.19: 48470480-48470480 on Build GRCh38
rs778693146 Chr.19: 48470515-48470515 on Build GRCh38
rs374014127 Chr.19: 48470589-48470589 on Build GRCh38
rs897075325 Chr.19: 48470550-48470550 on Build GRCh38
rs368161948 Chr.19: 48470617-48470617 on Build GRCh38
rs756802473 Chr.19: 48470615-48470615 on Build GRCh38
rs753321871 Chr.19: 48470507-48470507 on Build GRCh38
rs763086689 Chr.19: 48470557-48470557 on Build GRCh38
rs761989067 Chr.19: 48470453-48470453 on Build GRCh38
rs368394445 Chr.19: 48470579-48470579 on Build GRCh38
rs745703600 Chr.19: 48470519-48470519 on Build GRCh38
rs953782631 Chr.19: 48470567-48470567 on Build GRCh38
rs369334200 Chr.19: 48470529-48470529 on Build GRCh38
rs777160740 Chr.19: 48470506-48470506 on Build GRCh38
rs765259016 Chr.19: 48470463-48470463 on Build GRCh38
rs374616805 Chr.19: 48470613-48470613 on Build GRCh38
rs749261678 Chr.19: 48470524-48470525 on Build GRCh38
rs141221874 Chr.19: 48470491-48470491 on Build GRCh38
rs763101707 Chr.19: 48470469-48470469 on Build GRCh38
rs147667770 Chr.19: 48470468-48470468 on Build GRCh38
rs746938433 Chr.19: 48470525-48470525 on Build GRCh38
rs775542794 Chr.19: 48470447-48470447 on Build GRCh38
rs945508381 Chr.19: 48470601-48470601 on Build GRCh38
rs758071044 Chr.19: 48470624-48470624 on Build GRCh38
rs750127655 Chr.19: 48470623-48470623 on Build GRCh38
rs766525402 Chr.19: 48470471-48470471 on Build GRCh38
rs779951620 Chr.19: 48470524-48470524 on Build GRCh38
rs756604483 Chr.19: 48470496-48470496 on Build GRCh38
rs986540274 Chr.19: 48470576-48470576 on Build GRCh38
rs745652246 Chr.19: 48470436-48470436 on Build GRCh38
rs994205842 Chr.19: 48470569-48470569 on Build GRCh38
rs768676322 Chr.19: 48470554-48470555 on Build GRCh38
rs771203722 Chr.19: 48470562-48470562 on Build GRCh38
rs371156202 Chr.19: 48470534-48470534 on Build GRCh38
rs1004032218 Chr.19: 48470514-48470514 on Build GRCh38
rs201450029 Chr.19: 48470508-48470508 on Build GRCh38
rs772190797 Chr.19: 48470443-48470443 on Build GRCh38
rs118155821 Chr.19: 48470521-48470521 on Build GRCh38
rs774399664 Chr.19: 48470615-48470616 on Build GRCh38
rs764657368 Chr.19: 48470591-48470591 on Build GRCh38
rs889647249 Chr.19: 48470512-48470512 on Build GRCh38
rs553747052 Chr.19: 48470493-48470493 on Build GRCh38
rs755062556 Chr.19: 48470578-48470578 on Build GRCh38
rs200643268 Chr.19: 48470568-48470568 on Build GRCh38

View Full Product Details