Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64270441-64270666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf56
Entrez Gene ID
115749
Gene Name
chromosome 12 open reading frame 56
Gene Aliases
-
UniGene
Hs.535190
Gene Chromosome Location
Chr.12: 64264762-64390580 on Build GRCh38
Transcripts
NM_001099676.2 NM_001170633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246427 c.1171A>C p.S391R Chr.12: 64270648-64270648 on Build GRCh38
405184 c.1153delG p.A385fs*6 Chr.12: 64270666-64270666 on Build GRCh38
1363591 c.1259G>T p.R420M Chr.12: 64270560-64270560 on Build GRCh38
6990270 c.1219A>C p.S407R Chr.12: 64270600-64270600 on Build GRCh38
4853398 c.1162C>G p.Q388E Chr.12: 64270657-64270657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887890489 Chr.12: 64270625-64270625 on Build GRCh38
rs775801850 Chr.12: 64270600-64270600 on Build GRCh38
rs765932215 Chr.12: 64270643-64270643 on Build GRCh38
rs766056715 Chr.12: 64270549-64270549 on Build GRCh38
rs764456027 Chr.12: 64270521-64270521 on Build GRCh38
rs780984645 Chr.12: 64270570-64270570 on Build GRCh38
rs753335510 Chr.12: 64270528-64270528 on Build GRCh38
rs371749635 Chr.12: 64270596-64270596 on Build GRCh38
rs762364630 Chr.12: 64270540-64270540 on Build GRCh38
rs754570294 Chr.12: 64270566-64270566 on Build GRCh38
rs924095199 Chr.12: 64270561-64270561 on Build GRCh38
rs750421781 Chr.12: 64270570-64270571 on Build GRCh38
rs201212935 Chr.12: 64270640-64270640 on Build GRCh38
rs182706895 Chr.12: 64270603-64270603 on Build GRCh38
rs758365563 Chr.12: 64270581-64270583 on Build GRCh38
rs749484643 Chr.12: 64270579-64270579 on Build GRCh38
rs752688260 Chr.12: 64270571-64270571 on Build GRCh38
rs771248359 Chr.12: 64270586-64270586 on Build GRCh38
rs989574078 Chr.12: 64270597-64270597 on Build GRCh38
rs751008276 Chr.12: 64270559-64270559 on Build GRCh38
rs202117720 Chr.12: 64270590-64270590 on Build GRCh38
rs367929330 Chr.12: 64270547-64270547 on Build GRCh38
rs745922973 Chr.12: 64270591-64270591 on Build GRCh38
rs375405180 Chr.12: 64270548-64270548 on Build GRCh38
rs755920139 Chr.12: 64270575-64270575 on Build GRCh38
rs562853556 Chr.12: 64270577-64270577 on Build GRCh38
rs948005217 Chr.12: 64270599-64270599 on Build GRCh38
rs978762209 Chr.12: 64270505-64270505 on Build GRCh38
rs761193073 Chr.12: 64270601-64270601 on Build GRCh38
rs760881106 Chr.12: 64270495-64270495 on Build GRCh38
rs61547812 Chr.12: 64270474-64270474 on Build GRCh38
rs768967091 Chr.12: 64270602-64270602 on Build GRCh38

View Full Product Details