Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43283128-43283399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf210
Entrez Gene ID
149466
Gene Name
chromosome 1 open reading frame 210
Gene Aliases
TEMP
UniGene
Hs.158963
Gene Chromosome Location
Chr.1: 43281865-43285840 on Build GRCh38
Transcripts
NM_001164829.1 NM_182517.2 XM_011540802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7292909 c.19+8A>T p.? Chr.1: 43283244-43283244 on Build GRCh38
7358293 c.5A>G p.N2S Chr.1: 43283266-43283266 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs111305387 Chr.1: 43283266-43283266 on Build GRCh38
rs1017333011 Chr.1: 43283199-43283199 on Build GRCh38
rs543137591 Chr.1: 43283331-43283331 on Build GRCh38
rs368601161 Chr.1: 43283268-43283268 on Build GRCh38
rs371712175 Chr.1: 43283337-43283337 on Build GRCh38
rs762318970 Chr.1: 43283301-43283301 on Build GRCh38
rs780030451 Chr.1: 43283234-43283234 on Build GRCh38
rs893953330 Chr.1: 43283283-43283283 on Build GRCh38
rs990646525 Chr.1: 43283342-43283342 on Build GRCh38
rs143735190 Chr.1: 43283260-43283260 on Build GRCh38
rs534929390 Chr.1: 43283245-43283245 on Build GRCh38
rs747046418 Chr.1: 43283238-43283238 on Build GRCh38
rs367878230 Chr.1: 43283190-43283190 on Build GRCh38
rs765721252 Chr.1: 43283309-43283309 on Build GRCh38
rs554752457 Chr.1: 43283246-43283246 on Build GRCh38
rs11554559 Chr.1: 43283315-43283315 on Build GRCh38
rs754312239 Chr.1: 43283203-43283203 on Build GRCh38
rs770832882 Chr.1: 43283262-43283262 on Build GRCh38
rs767139763 Chr.1: 43283153-43283153 on Build GRCh38
rs755173042 Chr.1: 43283241-43283241 on Build GRCh38
rs990683027 Chr.1: 43283184-43283184 on Build GRCh38
rs918293670 Chr.1: 43283239-43283239 on Build GRCh38
rs1039226890 Chr.1: 43283269-43283269 on Build GRCh38
rs775253791 Chr.1: 43283295-43283295 on Build GRCh38
rs959416950 Chr.1: 43283170-43283170 on Build GRCh38
rs371385725 Chr.1: 43283278-43283278 on Build GRCh38
rs771688695 Chr.1: 43283288-43283288 on Build GRCh38
rs928055152 Chr.1: 43283160-43283160 on Build GRCh38
rs765886233 Chr.1: 43283218-43283218 on Build GRCh38
rs769550848 Chr.1: 43283256-43283256 on Build GRCh38
rs559231611 Chr.1: 43283174-43283174 on Build GRCh38
rs565931482 Chr.1: 43283204-43283204 on Build GRCh38
rs1007067290 Chr.1: 43283194-43283194 on Build GRCh38
rs745647430 Chr.1: 43283203-43283230 on Build GRCh38
rs989143845 Chr.1: 43283222-43283222 on Build GRCh38
rs750944824 Chr.1: 43283221-43283221 on Build GRCh38
rs772049254 Chr.1: 43283318-43283320 on Build GRCh38

View Full Product Details