Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24291014-24291245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS1
Entrez Gene ID
115817
Gene Name
dehydrogenase/reductase 1
Gene Aliases
SDR19C1
UniGene
Hs.348350
Gene Chromosome Location
Chr.14: 24290598-24299833 on Build GRCh38
Transcripts
NM_001136050.2 NM_138452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386498 c.781C>A p.R261S Chr.14: 24291163-24291163 on Build GRCh38
5910829 c.749G>A p.G250D Chr.14: 24291195-24291195 on Build GRCh38
4878245 c.805G>A p.G269S Chr.14: 24291139-24291139 on Build GRCh38
7427466 c.759G>C p.L253L Chr.14: 24291185-24291185 on Build GRCh38
2033116 c.793C>T p.R265W Chr.14: 24291151-24291151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752733121 Chr.14: 24291151-24291151 on Build GRCh38
rs200636329 Chr.14: 24291140-24291140 on Build GRCh38
rs768752833 Chr.14: 24291215-24291215 on Build GRCh38
rs370794166 Chr.14: 24291224-24291224 on Build GRCh38
rs201101817 Chr.14: 24291166-24291166 on Build GRCh38
rs934971060 Chr.14: 24291126-24291126 on Build GRCh38
rs997121611 Chr.14: 24291098-24291098 on Build GRCh38
rs899539298 Chr.14: 24291093-24291093 on Build GRCh38
rs2295310 Chr.14: 24291049-24291049 on Build GRCh38
rs368276525 Chr.14: 24291223-24291223 on Build GRCh38
rs34586111 Chr.14: 24291155-24291155 on Build GRCh38
rs575472175 Chr.14: 24291092-24291092 on Build GRCh38
rs545198591 Chr.14: 24291072-24291072 on Build GRCh38
rs745400245 Chr.14: 24291173-24291173 on Build GRCh38
rs148282342 Chr.14: 24291150-24291150 on Build GRCh38
rs757965746 Chr.14: 24291174-24291174 on Build GRCh38
rs201880552 Chr.14: 24291104-24291104 on Build GRCh38
rs559917499 Chr.14: 24291163-24291163 on Build GRCh38
rs2295309 Chr.14: 24291167-24291167 on Build GRCh38
rs762131126 Chr.14: 24291130-24291130 on Build GRCh38
rs758955966 Chr.14: 24291142-24291142 on Build GRCh38
rs771764036 Chr.14: 24291073-24291074 on Build GRCh38
rs765442917 Chr.14: 24291138-24291138 on Build GRCh38
rs200040073 Chr.14: 24291179-24291179 on Build GRCh38
rs200839029 Chr.14: 24291124-24291124 on Build GRCh38
rs756913740 Chr.14: 24291165-24291165 on Build GRCh38
rs747486685 Chr.14: 24291102-24291102 on Build GRCh38
rs771914758 Chr.14: 24291211-24291212 on Build GRCh38
rs777130594 Chr.14: 24291129-24291129 on Build GRCh38
rs59742678 Chr.14: 24291071-24291071 on Build GRCh38
rs773189484 Chr.14: 24291139-24291139 on Build GRCh38
rs375012309 Chr.14: 24291206-24291206 on Build GRCh38
rs147405102 Chr.14: 24291185-24291185 on Build GRCh38
rs148881510 Chr.14: 24291077-24291077 on Build GRCh38
rs763940229 Chr.14: 24291162-24291162 on Build GRCh38

View Full Product Details