Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69268884-69269145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLCE
Entrez Gene ID
26035
Gene Name
glucuronic acid epimerase
Gene Aliases
HSEPI
UniGene
Hs.183006
Gene Chromosome Location
Chr.15: 69160555-69272209 on Build GRCh38
Transcripts
NM_001324092.1 NM_001324093.1 NM_001324094.1 NM_015554.2 XM_005254298.3 XM_017022073.1 XM_017022074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3503509 c.1695C>T p.F565F Chr.15: 69269085-69269085 on Build GRCh38
4398248 c.1570G>A p.E524K Chr.15: 69268960-69268960 on Build GRCh38
5039540 c.1600G>A p.A534T Chr.15: 69268990-69268990 on Build GRCh38
3370238 c.1740C>T p.T580T Chr.15: 69269130-69269130 on Build GRCh38
1678560 c.1618C>T p.R540C Chr.15: 69269008-69269008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370606638 Chr.15: 69268912-69268912 on Build GRCh38
rs374601683 Chr.15: 69268956-69268956 on Build GRCh38
rs771177712 Chr.15: 69268983-69268983 on Build GRCh38
rs757117300 Chr.15: 69269070-69269070 on Build GRCh38
rs936411625 Chr.15: 69269003-69269003 on Build GRCh38
rs1052657514 Chr.15: 69269010-69269010 on Build GRCh38
rs201978364 Chr.15: 69269007-69269007 on Build GRCh38
rs756992856 Chr.15: 69268941-69268941 on Build GRCh38
rs530064318 Chr.15: 69268980-69268980 on Build GRCh38
rs756060048 Chr.15: 69268921-69268921 on Build GRCh38
rs763870631 Chr.15: 69268932-69268932 on Build GRCh38
rs267604304 Chr.15: 69269050-69269050 on Build GRCh38
rs112901621 Chr.15: 69269079-69269079 on Build GRCh38
rs764974808 Chr.15: 69269074-69269074 on Build GRCh38
rs925086202 Chr.15: 69268966-69268966 on Build GRCh38
rs892630807 Chr.15: 69269035-69269035 on Build GRCh38
rs775551601 Chr.15: 69269014-69269014 on Build GRCh38
rs779161883 Chr.15: 69269069-69269069 on Build GRCh38
rs137875799 Chr.15: 69269086-69269086 on Build GRCh38
rs778680256 Chr.15: 69268952-69268952 on Build GRCh38
rs377258384 Chr.15: 69269008-69269008 on Build GRCh38
rs368338173 Chr.15: 69268981-69268981 on Build GRCh38
rs561055553 Chr.15: 69268968-69268968 on Build GRCh38
rs753700982 Chr.15: 69269064-69269064 on Build GRCh38
rs906182858 Chr.15: 69268979-69268979 on Build GRCh38
rs746581109 Chr.15: 69269098-69269098 on Build GRCh38
rs758134548 Chr.15: 69269090-69269090 on Build GRCh38
rs760739746 Chr.15: 69269018-69269018 on Build GRCh38
rs753734612 Chr.15: 69268936-69268936 on Build GRCh38
rs779781921 Chr.15: 69269097-69269097 on Build GRCh38
rs757592519 Chr.15: 69269009-69269009 on Build GRCh38
rs774672416 Chr.15: 69268986-69268986 on Build GRCh38
rs770424681 Chr.15: 69268976-69268976 on Build GRCh38
rs764154374 Chr.15: 69269045-69269045 on Build GRCh38
rs746459456 Chr.15: 69268972-69268972 on Build GRCh38
rs745487276 Chr.15: 69268953-69268953 on Build GRCh38
rs947735422 Chr.15: 69268938-69268938 on Build GRCh38
rs372237500 Chr.15: 69269004-69269004 on Build GRCh38
rs1052032379 Chr.15: 69269114-69269114 on Build GRCh38

View Full Product Details