Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37950037-37950282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPDR1
Entrez Gene ID
54749
Gene Name
ependymin related 1
Gene Aliases
EPDR, MERP-1, MERP1, UCC1
UniGene
Hs.563491
Gene Chromosome Location
Chr.7: 37920561-37951941 on Build GRCh38
Transcripts
NM_001242946.1 NM_001242948.1 NM_017549.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777120 c.871G>C p.D291H Chr.7: 37950232-37950232 on Build GRCh38
6895166 c.858C>T p.I286I Chr.7: 37950219-37950219 on Build GRCh38
3412020 c.879T>C p.Y293Y Chr.7: 37950240-37950240 on Build GRCh38
1089466 c.878A>G p.Y293C Chr.7: 37950239-37950239 on Build GRCh38
6514907 c.882T>A p.P294P Chr.7: 37950243-37950243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111676835 Chr.7: 37950113-37950113 on Build GRCh38
rs537046780 Chr.7: 37950146-37950146 on Build GRCh38
rs1001723316 Chr.7: 37950184-37950184 on Build GRCh38
rs571749837 Chr.7: 37950228-37950228 on Build GRCh38
rs191827848 Chr.7: 37950186-37950186 on Build GRCh38
rs780991410 Chr.7: 37950170-37950170 on Build GRCh38
rs552173458 Chr.7: 37950103-37950103 on Build GRCh38
rs755923186 Chr.7: 37950183-37950183 on Build GRCh38
rs904064844 Chr.7: 37950063-37950063 on Build GRCh38
rs775328126 Chr.7: 37950184-37950184 on Build GRCh38
rs200925563 Chr.7: 37950212-37950212 on Build GRCh38
rs951880187 Chr.7: 37950189-37950189 on Build GRCh38
rs1050625172 Chr.7: 37950061-37950061 on Build GRCh38
rs1450851 Chr.7: 37950173-37950173 on Build GRCh38
rs745677612 Chr.7: 37950171-37950171 on Build GRCh38
rs766294494 Chr.7: 37950183-37950184 on Build GRCh38
rs987297148 Chr.7: 37950079-37950079 on Build GRCh38
rs768217022 Chr.7: 37950221-37950221 on Build GRCh38
rs770878040 Chr.7: 37950235-37950235 on Build GRCh38
rs777022716 Chr.7: 37950245-37950245 on Build GRCh38
rs761199728 Chr.7: 37950237-37950237 on Build GRCh38
rs532454317 Chr.7: 37950075-37950075 on Build GRCh38
rs1024452342 Chr.7: 37950160-37950160 on Build GRCh38
rs753743504 Chr.7: 37950190-37950190 on Build GRCh38
rs771376229 Chr.7: 37950239-37950239 on Build GRCh38
rs774014768 Chr.7: 37950233-37950233 on Build GRCh38
rs757154444 Chr.7: 37950169-37950169 on Build GRCh38
rs891451347 Chr.7: 37950123-37950123 on Build GRCh38

View Full Product Details