Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101245215-101245486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRP2
Entrez Gene ID
1821
Gene Name
dystrophin related protein 2
Gene Aliases
DRP-2
UniGene
Hs.159291
Gene Chromosome Location
Chr.X: 101219786-101264497 on Build GRCh38
Transcripts
NM_001171184.1 NM_001939.2 XM_017029333.1 XM_017029334.1

SNP Detail

SNP ID Chromosome Location
rs905013517 Chr.X: 101245246-101245246 on Build GRCh38
rs766695943 Chr.X: 101245343-101245343 on Build GRCh38
rs763727945 Chr.X: 101245406-101245406 on Build GRCh38
rs184611882 Chr.X: 101245318-101245318 on Build GRCh38
rs762654516 Chr.X: 101245400-101245400 on Build GRCh38
rs751069050 Chr.X: 101245441-101245441 on Build GRCh38
rs767509798 Chr.X: 101245458-101245458 on Build GRCh38
rs952660029 Chr.X: 101245380-101245380 on Build GRCh38
rs1028359815 Chr.X: 101245339-101245339 on Build GRCh38
rs757718978 Chr.X: 101245275-101245275 on Build GRCh38
rs750286160 Chr.X: 101245459-101245459 on Build GRCh38
rs1002056220 Chr.X: 101245303-101245303 on Build GRCh38
rs761167889 Chr.X: 101245443-101245443 on Build GRCh38
rs770382830 Chr.X: 101245338-101245341 on Build GRCh38
rs776961888 Chr.X: 101245368-101245368 on Build GRCh38

View Full Product Details