Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74205611-74205878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGT
Entrez Gene ID
8790
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
GFPP
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74208702 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Gene Symbol
FPGT-TNNI3K
Entrez Gene ID
100526835
Gene Name
FPGT-TNNI3K readthrough
Gene Aliases
-
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74544432 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
244594 c.1577G>A p.R526H Chr.1: 74205624-74205624 on Build GRCh38
1185487 c.1739G>T p.R580M Chr.1: 74205786-74205786 on Build GRCh38
4469850 c.1622C>T p.S541F Chr.1: 74205669-74205669 on Build GRCh38
1665476 c.1690delA p.I564fs*10 Chr.1: 74205737-74205737 on Build GRCh38
4652867 c.1762A>G p.S588G Chr.1: 74205809-74205809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774315921 Chr.1: 74205790-74205790 on Build GRCh38
rs768537296 Chr.1: 74205789-74205789 on Build GRCh38
rs190585230 Chr.1: 74205711-74205711 on Build GRCh38
rs754270559 Chr.1: 74205828-74205828 on Build GRCh38
rs775272386 Chr.1: 74205784-74205784 on Build GRCh38
rs778100815 Chr.1: 74205792-74205793 on Build GRCh38
rs372383046 Chr.1: 74205723-74205723 on Build GRCh38
rs767286333 Chr.1: 74205809-74205809 on Build GRCh38
rs755511609 Chr.1: 74205829-74205829 on Build GRCh38
rs767094085 Chr.1: 74205693-74205693 on Build GRCh38
rs748291587 Chr.1: 74205648-74205651 on Build GRCh38
rs760095046 Chr.1: 74205825-74205825 on Build GRCh38
rs910436473 Chr.1: 74205696-74205696 on Build GRCh38
rs749969984 Chr.1: 74205699-74205699 on Build GRCh38
rs749292769 Chr.1: 74205787-74205787 on Build GRCh38
rs943195384 Chr.1: 74205778-74205778 on Build GRCh38
rs368831037 Chr.1: 74205638-74205638 on Build GRCh38
rs562800576 Chr.1: 74205772-74205772 on Build GRCh38
rs757258443 Chr.1: 74205851-74205851 on Build GRCh38
rs769630889 Chr.1: 74205660-74205660 on Build GRCh38
rs777770818 Chr.1: 74205747-74205747 on Build GRCh38
rs780042474 Chr.1: 74205807-74205807 on Build GRCh38
rs761595383 Chr.1: 74205688-74205688 on Build GRCh38
rs1024833304 Chr.1: 74205687-74205687 on Build GRCh38
rs571848 Chr.1: 74205835-74205835 on Build GRCh38
rs779291014 Chr.1: 74205730-74205730 on Build GRCh38
rs756359056 Chr.1: 74205632-74205632 on Build GRCh38
rs377299844 Chr.1: 74205651-74205651 on Build GRCh38
rs748331899 Chr.1: 74205731-74205731 on Build GRCh38
rs535570007 Chr.1: 74205837-74205837 on Build GRCh38
rs371947111 Chr.1: 74205821-74205821 on Build GRCh38
rs148017212 Chr.1: 74205642-74205642 on Build GRCh38
rs759467099 Chr.1: 74205658-74205658 on Build GRCh38
rs772109404 Chr.1: 74205650-74205650 on Build GRCh38
rs762651188 Chr.1: 74205670-74205670 on Build GRCh38
rs978566745 Chr.1: 74205805-74205805 on Build GRCh38
rs764035240 Chr.1: 74205674-74205674 on Build GRCh38
rs766795952 Chr.1: 74205712-74205712 on Build GRCh38
rs775288139 Chr.1: 74205661-74205661 on Build GRCh38
rs372918657 Chr.1: 74205799-74205800 on Build GRCh38
rs551679590 Chr.1: 74205633-74205633 on Build GRCh38
rs373519356 Chr.1: 74205649-74205649 on Build GRCh38
rs779094163 Chr.1: 74205647-74205647 on Build GRCh38
rs771108793 Chr.1: 74205774-74205774 on Build GRCh38
rs777967231 Chr.1: 74205838-74205838 on Build GRCh38
rs747313963 Chr.1: 74205848-74205848 on Build GRCh38
rs774121145 Chr.1: 74205682-74205682 on Build GRCh38
rs531014346 Chr.1: 74205722-74205722 on Build GRCh38
rs747063121 Chr.1: 74205773-74205773 on Build GRCh38
rs781436495 Chr.1: 74205640-74205642 on Build GRCh38
rs758640235 Chr.1: 74205737-74205737 on Build GRCh38
rs765963504 Chr.1: 74205826-74205826 on Build GRCh38
rs756553679 Chr.1: 74205678-74205687 on Build GRCh38

View Full Product Details