Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93074175-93074420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP26A1
Entrez Gene ID
1592
Gene Name
cytochrome P450 family 26 subfamily A member 1
Gene Aliases
CP26, CYP26, P450RAI, P450RAI1
UniGene
Hs.150595
Gene Chromosome Location
Chr.10: 93073475-93077884 on Build GRCh38
Transcripts
NM_000783.3 NM_057157.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4758954 c.280G>A p.A94T Chr.10: 93074398-93074398 on Build GRCh38
4016668 c.85C>T p.R29W Chr.10: 93074410-93074410 on Build GRCh38
4016667 c.292C>T p.R98W Chr.10: 93074410-93074410 on Build GRCh38
7264048 c.296G>A p.R99H Chr.10: 93074414-93074414 on Build GRCh38
4016666 c.190C>T p.R64W Chr.10: 93074308-93074308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140561960 Chr.10: 93074340-93074340 on Build GRCh38
rs932316345 Chr.10: 93074197-93074197 on Build GRCh38
rs376693066 Chr.10: 93074273-93074273 on Build GRCh38
rs904422154 Chr.10: 93074225-93074225 on Build GRCh38
rs79228650 Chr.10: 93074264-93074264 on Build GRCh38
rs572000694 Chr.10: 93074220-93074220 on Build GRCh38
rs767835603 Chr.10: 93074388-93074388 on Build GRCh38
rs920914524 Chr.10: 93074194-93074194 on Build GRCh38
rs376312852 Chr.10: 93074325-93074325 on Build GRCh38
rs375269639 Chr.10: 93074252-93074252 on Build GRCh38
rs767299392 Chr.10: 93074266-93074266 on Build GRCh38
rs774249618 Chr.10: 93074262-93074262 on Build GRCh38
rs749344347 Chr.10: 93074313-93074313 on Build GRCh38
rs750057672 Chr.10: 93074382-93074382 on Build GRCh38
rs764756285 Chr.10: 93074374-93074374 on Build GRCh38
rs760754999 Chr.10: 93074354-93074354 on Build GRCh38
rs760627948 Chr.10: 93074270-93074270 on Build GRCh38
rs761442477 Chr.10: 93074373-93074373 on Build GRCh38
rs931420533 Chr.10: 93074200-93074200 on Build GRCh38
rs756607476 Chr.10: 93074302-93074302 on Build GRCh38
rs540974963 Chr.10: 93074253-93074253 on Build GRCh38
rs768695011 Chr.10: 93074371-93074371 on Build GRCh38
rs36098214 Chr.10: 93074262-93074263 on Build GRCh38
rs1046693823 Chr.10: 93074256-93074256 on Build GRCh38
rs185421897 Chr.10: 93074303-93074303 on Build GRCh38
rs764767781 Chr.10: 93074278-93074278 on Build GRCh38
rs934602790 Chr.10: 93074255-93074255 on Build GRCh38
rs551690765 Chr.10: 93074349-93074349 on Build GRCh38
rs757020975 Chr.10: 93074277-93074277 on Build GRCh38
rs750052606 Chr.10: 93074279-93074279 on Build GRCh38
rs1011103698 Chr.10: 93074271-93074271 on Build GRCh38
rs984144791 Chr.10: 93074386-93074386 on Build GRCh38
rs762599485 Chr.10: 93074387-93074387 on Build GRCh38
rs374194944 Chr.10: 93074269-93074269 on Build GRCh38
rs771126127 Chr.10: 93074260-93074260 on Build GRCh38
rs1042771925 Chr.10: 93074203-93074203 on Build GRCh38
rs369990701 Chr.10: 93074301-93074301 on Build GRCh38
rs776322111 Chr.10: 93074372-93074372 on Build GRCh38
rs1027638218 Chr.10: 93074364-93074364 on Build GRCh38
rs755322246 Chr.10: 93074292-93074292 on Build GRCh38
rs562861556 Chr.10: 93074333-93074333 on Build GRCh38

View Full Product Details