Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72008172-72008433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST3
Entrez Gene ID
9469
Gene Name
carbohydrate sulfotransferase 3
Gene Aliases
C6ST, C6ST1, HSD
UniGene
Hs.158304
Gene Chromosome Location
Chr.10: 71964362-72013564 on Build GRCh38
Transcripts
NM_004273.4 XM_006718075.3 XM_011540369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5590211 c.1164C>T p.F388F Chr.10: 72008195-72008195 on Build GRCh38
5499073 c.1297C>T p.P433S Chr.10: 72008328-72008328 on Build GRCh38
6425546 c.1298C>A p.P433H Chr.10: 72008329-72008329 on Build GRCh38
6368387 c.1328C>A p.A443D Chr.10: 72008359-72008359 on Build GRCh38
6396931 c.1192G>T p.E398* Chr.10: 72008223-72008223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756147851 Chr.10: 72008215-72008215 on Build GRCh38
rs774866759 Chr.10: 72008293-72008293 on Build GRCh38
rs1035424190 Chr.10: 72008200-72008200 on Build GRCh38
rs761600756 Chr.10: 72008356-72008356 on Build GRCh38
rs866564371 Chr.10: 72008251-72008251 on Build GRCh38
rs547001828 Chr.10: 72008213-72008213 on Build GRCh38
rs867758030 Chr.10: 72008344-72008344 on Build GRCh38
rs1004650270 Chr.10: 72008273-72008273 on Build GRCh38
rs965883985 Chr.10: 72008329-72008329 on Build GRCh38
rs200249458 Chr.10: 72008378-72008378 on Build GRCh38
rs758296485 Chr.10: 72008374-72008374 on Build GRCh38
rs771572867 Chr.10: 72008296-72008296 on Build GRCh38
rs951331805 Chr.10: 72008245-72008245 on Build GRCh38
rs76803307 Chr.10: 72008384-72008384 on Build GRCh38
rs561366883 Chr.10: 72008195-72008195 on Build GRCh38
rs779115203 Chr.10: 72008333-72008333 on Build GRCh38
rs905033055 Chr.10: 72008218-72008218 on Build GRCh38
rs184636110 Chr.10: 72008228-72008228 on Build GRCh38
rs771208378 Chr.10: 72008292-72008292 on Build GRCh38
rs1003539672 Chr.10: 72008196-72008196 on Build GRCh38
rs975600744 Chr.10: 72008330-72008330 on Build GRCh38
rs750324249 Chr.10: 72008365-72008365 on Build GRCh38
rs763898385 Chr.10: 72008345-72008345 on Build GRCh38
rs374954414 Chr.10: 72008394-72008394 on Build GRCh38
rs115579748 Chr.10: 72008204-72008204 on Build GRCh38
rs748327640 Chr.10: 72008259-72008259 on Build GRCh38
rs988768654 Chr.10: 72008372-72008372 on Build GRCh38
rs886042739 Chr.10: 72008222-72008222 on Build GRCh38
rs1052557339 Chr.10: 72008401-72008401 on Build GRCh38
rs547960656 Chr.10: 72008270-72008270 on Build GRCh38
rs985887467 Chr.10: 72008287-72008287 on Build GRCh38
rs773733746 Chr.10: 72008193-72008193 on Build GRCh38
rs939425272 Chr.10: 72008412-72008412 on Build GRCh38
rs767987808 Chr.10: 72008291-72008293 on Build GRCh38
rs145384892 Chr.10: 72008191-72008191 on Build GRCh38
rs749724374 Chr.10: 72008291-72008291 on Build GRCh38
rs764953708 Chr.10: 72008358-72008358 on Build GRCh38
rs760437804 Chr.10: 72008205-72008205 on Build GRCh38
rs778084595 Chr.10: 72008285-72008285 on Build GRCh38
rs755025272 Chr.10: 72008238-72008238 on Build GRCh38
rs763951694 Chr.10: 72008211-72008211 on Build GRCh38
rs766952355 Chr.10: 72008197-72008197 on Build GRCh38
rs756311245 Chr.10: 72008382-72008382 on Build GRCh38
rs1013403968 Chr.10: 72008206-72008206 on Build GRCh38
rs993806926 Chr.10: 72008214-72008214 on Build GRCh38
rs745384935 Chr.10: 72008366-72008366 on Build GRCh38
rs140411224 Chr.10: 72008282-72008282 on Build GRCh38
rs551353626 Chr.10: 72008410-72008410 on Build GRCh38
rs752819533 Chr.10: 72008381-72008381 on Build GRCh38
rs369055984 Chr.10: 72008411-72008411 on Build GRCh38
rs776562348 Chr.10: 72008355-72008355 on Build GRCh38
rs757595872 Chr.10: 72008407-72008407 on Build GRCh38
rs946848455 Chr.10: 72008386-72008386 on Build GRCh38
rs749488516 Chr.10: 72008396-72008396 on Build GRCh38
rs866792272 Chr.10: 72008409-72008409 on Build GRCh38
rs995382252 Chr.10: 72008221-72008221 on Build GRCh38
rs372291815 Chr.10: 72008297-72008297 on Build GRCh38
rs746356934 Chr.10: 72008414-72008414 on Build GRCh38
rs781152270 Chr.10: 72008239-72008239 on Build GRCh38
rs746386664 Chr.10: 72008294-72008294 on Build GRCh38

View Full Product Details