Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 92924076-92924349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD2
Entrez Gene ID
1106
Gene Name
chromodomain helicase DNA binding protein 2
Gene Aliases
EEOC
UniGene
Hs.220864
Gene Chromosome Location
Chr.15: 92900321-93028007 on Build GRCh38
Transcripts
NM_001042572.2 NM_001271.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
702081 c.87G>T p.S29S Chr.15: 92924345-92924345 on Build GRCh38
966688 c.87G>A p.S29S Chr.15: 92924345-92924345 on Build GRCh38
5573511 c.63-1G>A p.? Chr.15: 92924320-92924320 on Build GRCh38
702080 c.87G>T p.S29S Chr.15: 92924345-92924345 on Build GRCh38
7175247 c.68C>T p.S23L Chr.15: 92924326-92924326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979509149 Chr.15: 92924171-92924171 on Build GRCh38
rs1030703514 Chr.15: 92924201-92924201 on Build GRCh38
rs554333943 Chr.15: 92924113-92924113 on Build GRCh38
rs535294505 Chr.15: 92924114-92924114 on Build GRCh38
rs564938079 Chr.15: 92924162-92924162 on Build GRCh38
rs1018057756 Chr.15: 92924134-92924134 on Build GRCh38
rs992116960 Chr.15: 92924228-92924228 on Build GRCh38
rs8026695 Chr.15: 92924124-92924124 on Build GRCh38
rs766960119 Chr.15: 92924300-92924300 on Build GRCh38
rs766069247 Chr.15: 92924299-92924300 on Build GRCh38
rs148969999 Chr.15: 92924261-92924261 on Build GRCh38
rs947907952 Chr.15: 92924245-92924245 on Build GRCh38
rs946437117 Chr.15: 92924225-92924225 on Build GRCh38
rs544204390 Chr.15: 92924235-92924235 on Build GRCh38
rs373817674 Chr.15: 92924253-92924253 on Build GRCh38
rs1030321620 Chr.15: 92924117-92924117 on Build GRCh38
rs998482476 Chr.15: 92924250-92924250 on Build GRCh38
rs1007047545 Chr.15: 92924128-92924128 on Build GRCh38
rs780395101 Chr.15: 92924184-92924184 on Build GRCh38
rs753257921 Chr.15: 92924302-92924302 on Build GRCh38
rs546536259 Chr.15: 92924271-92924271 on Build GRCh38
rs937831429 Chr.15: 92924170-92924170 on Build GRCh38
rs749583665 Chr.15: 92924299-92924302 on Build GRCh38
rs530556215 Chr.15: 92924212-92924212 on Build GRCh38
rs372471036 Chr.15: 92924306-92924306 on Build GRCh38
rs77734149 Chr.15: 92924107-92924107 on Build GRCh38
rs1054933589 Chr.15: 92924192-92924192 on Build GRCh38
rs529988849 Chr.15: 92924265-92924265 on Build GRCh38
rs3826036 Chr.15: 92924161-92924161 on Build GRCh38
rs563636111 Chr.15: 92924246-92924246 on Build GRCh38
rs1051284586 Chr.15: 92924256-92924256 on Build GRCh38

View Full Product Details