Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125723752-125723987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDRF1
Entrez Gene ID
26098
Gene Name
erythroid differentiation regulatory factor 1
Gene Aliases
C10orf137
UniGene
Hs.468688
Gene Chromosome Location
Chr.10: 125719515-125764143 on Build GRCh38
Transcripts
NM_001202438.1 NM_015608.2 NR_110857.1 NR_110858.1 NR_110859.1 XM_017016066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2021512 c.445C>T p.R149C Chr.10: 125723871-125723871 on Build GRCh38
2021516 c.506C>T p.S169F Chr.10: 125723932-125723932 on Build GRCh38
2021514 c.446G>A p.R149H Chr.10: 125723872-125723872 on Build GRCh38
126574 c.508C>T p.Q170* Chr.10: 125723934-125723934 on Build GRCh38
2021511 c.445C>T p.R149C Chr.10: 125723871-125723871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942360785 Chr.10: 125723921-125723921 on Build GRCh38
rs765469318 Chr.10: 125723943-125723943 on Build GRCh38
rs781065462 Chr.10: 125723797-125723797 on Build GRCh38
rs910926164 Chr.10: 125723918-125723918 on Build GRCh38
rs572379699 Chr.10: 125723800-125723800 on Build GRCh38
rs866235128 Chr.10: 125723836-125723836 on Build GRCh38
rs372174581 Chr.10: 125723788-125723788 on Build GRCh38
rs759711447 Chr.10: 125723825-125723825 on Build GRCh38
rs754084555 Chr.10: 125723870-125723870 on Build GRCh38
rs778391588 Chr.10: 125723927-125723927 on Build GRCh38
rs200081757 Chr.10: 125723807-125723807 on Build GRCh38
rs779907121 Chr.10: 125723907-125723907 on Build GRCh38
rs765326292 Chr.10: 125723872-125723872 on Build GRCh38
rs553992713 Chr.10: 125723790-125723790 on Build GRCh38
rs755239860 Chr.10: 125723871-125723871 on Build GRCh38
rs919293608 Chr.10: 125723789-125723789 on Build GRCh38
rs748969247 Chr.10: 125723814-125723814 on Build GRCh38
rs768379204 Chr.10: 125723912-125723912 on Build GRCh38
rs766846778 Chr.10: 125723864-125723864 on Build GRCh38
rs761170508 Chr.10: 125723853-125723853 on Build GRCh38
rs757206079 Chr.10: 125723791-125723791 on Build GRCh38
rs565173559 Chr.10: 125723886-125723886 on Build GRCh38
rs754232535 Chr.10: 125723950-125723951 on Build GRCh38
rs771687542 Chr.10: 125723930-125723930 on Build GRCh38
rs922427664 Chr.10: 125723882-125723882 on Build GRCh38
rs745787655 Chr.10: 125723900-125723900 on Build GRCh38
rs773928359 Chr.10: 125723831-125723831 on Build GRCh38
rs748980610 Chr.10: 125723911-125723911 on Build GRCh38
rs752851632 Chr.10: 125723874-125723874 on Build GRCh38
rs909762378 Chr.10: 125723809-125723809 on Build GRCh38
rs374068896 Chr.10: 125723929-125723929 on Build GRCh38
rs367690424 Chr.10: 125723783-125723783 on Build GRCh38
rs773825217 Chr.10: 125723852-125723852 on Build GRCh38
rs61738566 Chr.10: 125723903-125723903 on Build GRCh38
rs754069735 Chr.10: 125723786-125723786 on Build GRCh38
rs761181970 Chr.10: 125723849-125723849 on Build GRCh38
rs758438991 Chr.10: 125723884-125723884 on Build GRCh38

View Full Product Details