Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38693436-38693703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE8
Entrez Gene ID
144402
Gene Name
copine 8
Gene Aliases
-
UniGene
Hs.40910
Gene Chromosome Location
Chr.12: 38652200-38908593 on Build GRCh38
Transcripts
NM_153634.2 XM_011537951.2 XM_017018851.1 XM_017018852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634705 c.1143G>A p.L381L Chr.12: 38693657-38693657 on Build GRCh38
3871520 c.1112C>T p.P371L Chr.12: 38693688-38693688 on Build GRCh38
1361307 c.1132G>A p.E378K Chr.12: 38693668-38693668 on Build GRCh38
1361306 c.1142T>A p.L381* Chr.12: 38693658-38693658 on Build GRCh38
5438876 c.1118G>T p.G373V Chr.12: 38693682-38693682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778610634 Chr.12: 38693622-38693622 on Build GRCh38
rs1053129899 Chr.12: 38693502-38693502 on Build GRCh38
rs933363475 Chr.12: 38693487-38693487 on Build GRCh38
rs534027928 Chr.12: 38693534-38693534 on Build GRCh38
rs776868973 Chr.12: 38693678-38693678 on Build GRCh38
rs778877068 Chr.12: 38693627-38693627 on Build GRCh38
rs996327829 Chr.12: 38693640-38693640 on Build GRCh38
rs973028399 Chr.12: 38693642-38693642 on Build GRCh38
rs143098149 Chr.12: 38693486-38693486 on Build GRCh38
rs757164876 Chr.12: 38693638-38693638 on Build GRCh38
rs1028819232 Chr.12: 38693649-38693649 on Build GRCh38
rs878949568 Chr.12: 38693574-38693574 on Build GRCh38
rs1047304665 Chr.12: 38693563-38693563 on Build GRCh38
rs201121630 Chr.12: 38693669-38693669 on Build GRCh38
rs745635574 Chr.12: 38693646-38693646 on Build GRCh38
rs560423315 Chr.12: 38693581-38693581 on Build GRCh38
rs1011678898 Chr.12: 38693511-38693511 on Build GRCh38
rs529115632 Chr.12: 38693610-38693610 on Build GRCh38
rs1046984886 Chr.12: 38693522-38693532 on Build GRCh38
rs940338551 Chr.12: 38693590-38693590 on Build GRCh38
rs540030759 Chr.12: 38693549-38693549 on Build GRCh38
rs1025322893 Chr.12: 38693662-38693662 on Build GRCh38
rs772563288 Chr.12: 38693623-38693623 on Build GRCh38
rs747521278 Chr.12: 38693668-38693668 on Build GRCh38
rs573428539 Chr.12: 38693464-38693464 on Build GRCh38
rs574970931 Chr.12: 38693550-38693550 on Build GRCh38
rs370670405 Chr.12: 38693615-38693615 on Build GRCh38
rs577914128 Chr.12: 38693509-38693509 on Build GRCh38
rs886122387 Chr.12: 38693570-38693570 on Build GRCh38
rs1039290354 Chr.12: 38693462-38693462 on Build GRCh38

View Full Product Details