Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118898927-118899156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL9L
Entrez Gene ID
283149
Gene Name
B-cell CLL/lymphoma 9-like
Gene Aliases
BCL9-2, DLNB11
UniGene
Hs.414740
Gene Chromosome Location
Chr.11: 118896140-118926072 on Build GRCh38
Transcripts
NM_182557.2 XM_005271511.2 XM_005271512.2 XM_005271513.2 XM_006718815.2 XM_011542756.2 XM_017017572.1 XM_017017573.1 XM_017017574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1351720 c.3956C>T p.T1319M Chr.11: 118898959-118898959 on Build GRCh38
4665458 c.3972C>T p.F1324F Chr.11: 118898943-118898943 on Build GRCh38
3383238 c.3894C>A p.L1298L Chr.11: 118899021-118899021 on Build GRCh38
3808513 c.3938T>G p.V1313G Chr.11: 118898977-118898977 on Build GRCh38
6858392 c.3887G>A p.G1296D Chr.11: 118899028-118899028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748594743 Chr.11: 118898958-118898958 on Build GRCh38
rs777767215 Chr.11: 118899031-118899031 on Build GRCh38
rs1034028465 Chr.11: 118898993-118898993 on Build GRCh38
rs776370296 Chr.11: 118899062-118899062 on Build GRCh38
rs542302053 Chr.11: 118899111-118899111 on Build GRCh38
rs762517662 Chr.11: 118899091-118899091 on Build GRCh38
rs751266481 Chr.11: 118899094-118899094 on Build GRCh38
rs765197776 Chr.11: 118899078-118899078 on Build GRCh38
rs749377217 Chr.11: 118899040-118899040 on Build GRCh38
rs182557494 Chr.11: 118898949-118898949 on Build GRCh38
rs199836573 Chr.11: 118898990-118898990 on Build GRCh38
rs779741410 Chr.11: 118898996-118898996 on Build GRCh38
rs933323171 Chr.11: 118899102-118899102 on Build GRCh38
rs761689835 Chr.11: 118898981-118898981 on Build GRCh38
rs773305789 Chr.11: 118898960-118898960 on Build GRCh38
rs141872454 Chr.11: 118899042-118899042 on Build GRCh38
rs200803594 Chr.11: 118899126-118899126 on Build GRCh38
rs150852065 Chr.11: 118898959-118898959 on Build GRCh38
rs749957297 Chr.11: 118899080-118899080 on Build GRCh38
rs919223513 Chr.11: 118899098-118899098 on Build GRCh38
rs753786456 Chr.11: 118898979-118898979 on Build GRCh38
rs763128397 Chr.11: 118898962-118898962 on Build GRCh38
rs139252145 Chr.11: 118898997-118898997 on Build GRCh38
rs370572120 Chr.11: 118899105-118899105 on Build GRCh38
rs933964892 Chr.11: 118899026-118899026 on Build GRCh38
rs754641285 Chr.11: 118899097-118899097 on Build GRCh38
rs759697112 Chr.11: 118899045-118899045 on Build GRCh38
rs755394615 Chr.11: 118899003-118899003 on Build GRCh38
rs143065470 Chr.11: 118898948-118898948 on Build GRCh38
rs776636657 Chr.11: 118899030-118899030 on Build GRCh38
rs566574462 Chr.11: 118899053-118899053 on Build GRCh38
rs560556908 Chr.11: 118899117-118899117 on Build GRCh38
rs760459717 Chr.11: 118898978-118898978 on Build GRCh38
rs562643900 Chr.11: 118899021-118899021 on Build GRCh38
rs547958566 Chr.11: 118898991-118898991 on Build GRCh38
rs766604306 Chr.11: 118898963-118898963 on Build GRCh38
rs768461855 Chr.11: 118899054-118899054 on Build GRCh38
rs771126846 Chr.11: 118899041-118899041 on Build GRCh38
rs770307487 Chr.11: 118899028-118899028 on Build GRCh38
rs200223163 Chr.11: 118898982-118898982 on Build GRCh38
rs761695075 Chr.11: 118899072-118899072 on Build GRCh38
rs375444038 Chr.11: 118899119-118899119 on Build GRCh38
rs775208557 Chr.11: 118898972-118898972 on Build GRCh38
rs781639270 Chr.11: 118899016-118899016 on Build GRCh38
rs765736563 Chr.11: 118899093-118899093 on Build GRCh38

View Full Product Details