Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180553343-180553568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT6
Entrez Gene ID
57472
Gene Name
CCR4-NOT transcription complex subunit 6
Gene Aliases
CCR4, Ccr4a
UniGene
Hs.608380
Gene Chromosome Location
Chr.5: 180494373-180578405 on Build GRCh38
Transcripts
NM_001303241.1 XM_005265953.1 XM_011534605.2 XM_011534606.1 XM_011534607.1 XM_017009670.1 XM_017009671.1 XM_017009672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6490692 c.300-9_300-7delCAT p.? Chr.5: 180553377-180553379 on Build GRCh38
1066792 c.328C>T p.R110* Chr.5: 180553414-180553414 on Build GRCh38
6631534 c.343G>C p.E115Q Chr.5: 180553429-180553429 on Build GRCh38
6470238 c.385+4T>G p.? Chr.5: 180553475-180553475 on Build GRCh38
5788683 c.328C>A p.R110R Chr.5: 180553414-180553414 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754633058 Chr.5: 180553489-180553489 on Build GRCh38
rs757148073 Chr.5: 180553508-180553509 on Build GRCh38
rs753537874 Chr.5: 180553454-180553454 on Build GRCh38
rs751352753 Chr.5: 180553503-180553504 on Build GRCh38
rs775657696 Chr.5: 180553422-180553422 on Build GRCh38
rs765028076 Chr.5: 180553460-180553460 on Build GRCh38
rs774500626 Chr.5: 180553415-180553415 on Build GRCh38
rs954737570 Chr.5: 180553526-180553526 on Build GRCh38
rs751238230 Chr.5: 180553483-180553483 on Build GRCh38
rs560086983 Chr.5: 180553379-180553379 on Build GRCh38
rs11738129 Chr.5: 180553527-180553527 on Build GRCh38
rs552235141 Chr.5: 180553474-180553474 on Build GRCh38
rs72814958 Chr.5: 180553378-180553378 on Build GRCh38
rs761815419 Chr.5: 180553457-180553457 on Build GRCh38
rs749828356 Chr.5: 180553388-180553388 on Build GRCh38
rs779841468 Chr.5: 180553376-180553376 on Build GRCh38
rs778200069 Chr.5: 180553491-180553491 on Build GRCh38
rs771272017 Chr.5: 180553414-180553414 on Build GRCh38
rs894559784 Chr.5: 180553478-180553478 on Build GRCh38
rs903086230 Chr.5: 180553479-180553479 on Build GRCh38
rs532397227 Chr.5: 180553382-180553382 on Build GRCh38
rs749554003 Chr.5: 180553494-180553494 on Build GRCh38
rs199907095 Chr.5: 180553464-180553464 on Build GRCh38
rs759518869 Chr.5: 180553417-180553417 on Build GRCh38
rs757980994 Chr.5: 180553471-180553471 on Build GRCh38
rs1042940569 Chr.5: 180553475-180553475 on Build GRCh38
rs760512456 Chr.5: 180553424-180553424 on Build GRCh38
rs750009649 Chr.5: 180553462-180553462 on Build GRCh38
rs745986799 Chr.5: 180553523-180553523 on Build GRCh38
rs372696271 Chr.5: 180553446-180553446 on Build GRCh38
rs779281332 Chr.5: 180553515-180553515 on Build GRCh38
rs376940712 Chr.5: 180553509-180553509 on Build GRCh38

View Full Product Details