Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168714936-168715207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS6
Entrez Gene ID
253782
Gene Name
ceramide synthase 6
Gene Aliases
CERS5, LASS6
UniGene
Hs.506829
Gene Chromosome Location
Chr.2: 168456249-168775134 on Build GRCh38
Transcripts
NM_001256126.1 NM_203463.2 XM_005246440.4 XM_017003749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238058 c.686G>T p.R229L Chr.2: 168715077-168715077 on Build GRCh38
3569581 c.691G>A p.G231R Chr.2: 168715082-168715082 on Build GRCh38
4595784 c.730C>A p.L244I Chr.2: 168715121-168715121 on Build GRCh38
3569582 c.730C>T p.L244F Chr.2: 168715121-168715121 on Build GRCh38
7316832 c.654G>A p.L218L Chr.2: 168715045-168715045 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774014075 Chr.2: 168715079-168715079 on Build GRCh38
rs747608813 Chr.2: 168715181-168715181 on Build GRCh38
rs989821665 Chr.2: 168714987-168714987 on Build GRCh38
rs781249433 Chr.2: 168715071-168715071 on Build GRCh38
rs781470081 Chr.2: 168715145-168715145 on Build GRCh38
rs1032356843 Chr.2: 168715108-168715108 on Build GRCh38
rs779197751 Chr.2: 168714995-168714995 on Build GRCh38
rs780954291 Chr.2: 168715077-168715077 on Build GRCh38
rs756044038 Chr.2: 168714981-168714981 on Build GRCh38
rs1050247899 Chr.2: 168715095-168715095 on Build GRCh38
rs747182302 Chr.2: 168715010-168715010 on Build GRCh38
rs375567826 Chr.2: 168715166-168715166 on Build GRCh38
rs146657043 Chr.2: 168715087-168715087 on Build GRCh38
rs780260977 Chr.2: 168715121-168715121 on Build GRCh38
rs368211276 Chr.2: 168715038-168715038 on Build GRCh38
rs770077691 Chr.2: 168715161-168715161 on Build GRCh38
rs768953712 Chr.2: 168715024-168715024 on Build GRCh38
rs758488825 Chr.2: 168715000-168715000 on Build GRCh38
rs781591758 Chr.2: 168715064-168715064 on Build GRCh38
rs759203411 Chr.2: 168715085-168715085 on Build GRCh38
rs766235835 Chr.2: 168714997-168714997 on Build GRCh38
rs116506340 Chr.2: 168715009-168715009 on Build GRCh38
rs892017039 Chr.2: 168715136-168715136 on Build GRCh38
rs751771710 Chr.2: 168715122-168715122 on Build GRCh38
rs372323865 Chr.2: 168715076-168715076 on Build GRCh38
rs758577445 Chr.2: 168715112-168715112 on Build GRCh38
rs754814456 Chr.2: 168715065-168715065 on Build GRCh38
rs755225302 Chr.2: 168715135-168715135 on Build GRCh38
rs752622547 Chr.2: 168714977-168714977 on Build GRCh38
rs773651193 Chr.2: 168715073-168715073 on Build GRCh38
rs764137443 Chr.2: 168715098-168715098 on Build GRCh38
rs770321860 Chr.2: 168715066-168715066 on Build GRCh38
rs749744852 Chr.2: 168715168-168715168 on Build GRCh38
rs369910905 Chr.2: 168714965-168714965 on Build GRCh38
rs767291291 Chr.2: 168714971-168714971 on Build GRCh38
rs116510340 Chr.2: 168715100-168715100 on Build GRCh38
rs760583543 Chr.2: 168715096-168715096 on Build GRCh38
rs748560125 Chr.2: 168715149-168715149 on Build GRCh38
rs773816652 Chr.2: 168715176-168715176 on Build GRCh38

View Full Product Details