Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 69929428-69929685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL19A1
Entrez Gene ID
1310
Gene Name
collagen type XIX alpha 1 chain
Gene Aliases
COL9A1L, D6S228E
UniGene
Hs.444842
Gene Chromosome Location
Chr.6: 69866556-70215063 on Build GRCh38
Transcripts
NM_001858.5 XM_011535437.2 XM_011535438.2 XM_017010253.1 XM_017010254.1 XM_017010255.1 XM_017010256.1 XM_017010257.1 XM_017010258.1 XM_017010259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502522 c.615C>T p.F205F Chr.6: 69929649-69929649 on Build GRCh38
3875190 c.521T>A p.L174H Chr.6: 69929555-69929555 on Build GRCh38
3875191 c.592G>A p.D198N Chr.6: 69929626-69929626 on Build GRCh38
6632707 c.623G>A p.R208Q Chr.6: 69929657-69929657 on Build GRCh38
1697496 c.427G>A p.E143K Chr.6: 69929461-69929461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759924510 Chr.6: 69929612-69929612 on Build GRCh38
rs112596596 Chr.6: 69929527-69929527 on Build GRCh38
rs766867170 Chr.6: 69929592-69929592 on Build GRCh38
rs2229798 Chr.6: 69929657-69929657 on Build GRCh38
rs151153446 Chr.6: 69929516-69929516 on Build GRCh38
rs542927549 Chr.6: 69929636-69929636 on Build GRCh38
rs754835972 Chr.6: 69929638-69929638 on Build GRCh38
rs774739335 Chr.6: 69929605-69929605 on Build GRCh38
rs780113833 Chr.6: 69929629-69929629 on Build GRCh38
rs942718234 Chr.6: 69929550-69929550 on Build GRCh38
rs777797605 Chr.6: 69929637-69929637 on Build GRCh38
rs145098172 Chr.6: 69929568-69929568 on Build GRCh38
rs765185152 Chr.6: 69929484-69929484 on Build GRCh38
rs1016148358 Chr.6: 69929463-69929463 on Build GRCh38
rs776936776 Chr.6: 69929574-69929574 on Build GRCh38
rs778498718 Chr.6: 69929554-69929554 on Build GRCh38
rs780786675 Chr.6: 69929639-69929639 on Build GRCh38
rs550061144 Chr.6: 69929473-69929473 on Build GRCh38
rs748419367 Chr.6: 69929651-69929651 on Build GRCh38
rs763959008 Chr.6: 69929588-69929588 on Build GRCh38
rs142247604 Chr.6: 69929569-69929569 on Build GRCh38
rs763509709 Chr.6: 69929587-69929587 on Build GRCh38
rs761927803 Chr.6: 69929477-69929477 on Build GRCh38
rs748281349 Chr.6: 69929579-69929579 on Build GRCh38
rs954562339 Chr.6: 69929538-69929538 on Build GRCh38
rs569649456 Chr.6: 69929485-69929485 on Build GRCh38
rs199582099 Chr.6: 69929525-69929525 on Build GRCh38
rs772373887 Chr.6: 69929584-69929584 on Build GRCh38
rs753339843 Chr.6: 69929540-69929540 on Build GRCh38
rs186826494 Chr.6: 69929656-69929656 on Build GRCh38
rs146015062 Chr.6: 69929581-69929581 on Build GRCh38
rs866890954 Chr.6: 69929529-69929529 on Build GRCh38
rs200842660 Chr.6: 69929624-69929624 on Build GRCh38
rs756820427 Chr.6: 69929543-69929543 on Build GRCh38
rs745813614 Chr.6: 69929557-69929557 on Build GRCh38
rs768914724 Chr.6: 69929472-69929472 on Build GRCh38
rs763387613 Chr.6: 69929490-69929490 on Build GRCh38
rs756800886 Chr.6: 69929620-69929620 on Build GRCh38
rs773223199 Chr.6: 69929585-69929585 on Build GRCh38
rs150284256 Chr.6: 69929539-69929539 on Build GRCh38
rs746223283 Chr.6: 69929466-69929467 on Build GRCh38
rs760860605 Chr.6: 69929456-69929456 on Build GRCh38
rs138814921 Chr.6: 69929613-69929613 on Build GRCh38
rs573853099 Chr.6: 69929627-69929627 on Build GRCh38
rs375356627 Chr.6: 69929570-69929570 on Build GRCh38
rs866179157 Chr.6: 69929515-69929515 on Build GRCh38

View Full Product Details