Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161752271-161752530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP12
Entrez Gene ID
11266
Gene Name
dual specificity phosphatase 12
Gene Aliases
DUSP1, YVH1
UniGene
Hs.416216
Gene Chromosome Location
Chr.1: 161749692-161757164 on Build GRCh38
Transcripts
NM_007240.2 XM_005244862.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5978146 c.665G>C p.R222T Chr.1: 161752455-161752455 on Build GRCh38
3997033 c.624C>T p.T208T Chr.1: 161752414-161752414 on Build GRCh38
5844285 c.662G>A p.C221Y Chr.1: 161752452-161752452 on Build GRCh38
3477340 c.646G>A p.E216K Chr.1: 161752436-161752436 on Build GRCh38
6282883 c.601C>T p.L201F Chr.1: 161752391-161752391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371381196 Chr.1: 161752336-161752336 on Build GRCh38
rs1063179 Chr.1: 161752414-161752414 on Build GRCh38
rs771146440 Chr.1: 161752399-161752399 on Build GRCh38
rs1007671791 Chr.1: 161752324-161752324 on Build GRCh38
rs540206625 Chr.1: 161752483-161752483 on Build GRCh38
rs752542802 Chr.1: 161752434-161752434 on Build GRCh38
rs745399124 Chr.1: 161752488-161752489 on Build GRCh38
rs889362114 Chr.1: 161752303-161752303 on Build GRCh38
rs781572495 Chr.1: 161752361-161752361 on Build GRCh38
rs759631554 Chr.1: 161752415-161752415 on Build GRCh38
rs898795119 Chr.1: 161752342-161752342 on Build GRCh38
rs537560534 Chr.1: 161752387-161752387 on Build GRCh38
rs993215171 Chr.1: 161752352-161752352 on Build GRCh38
rs10917852 Chr.1: 161752482-161752482 on Build GRCh38
rs769246707 Chr.1: 161752345-161752345 on Build GRCh38
rs756656143 Chr.1: 161752495-161752495 on Build GRCh38
rs767681267 Chr.1: 161752422-161752422 on Build GRCh38
rs765014465 Chr.1: 161752465-161752465 on Build GRCh38
rs562175294 Chr.1: 161752472-161752472 on Build GRCh38
rs770024507 Chr.1: 161752385-161752385 on Build GRCh38
rs763786235 Chr.1: 161752455-161752455 on Build GRCh38
rs763101035 Chr.1: 161752390-161752390 on Build GRCh38
rs889736663 Chr.1: 161752403-161752403 on Build GRCh38
rs374983278 Chr.1: 161752456-161752456 on Build GRCh38
rs749904686 Chr.1: 161752467-161752467 on Build GRCh38
rs778637924 Chr.1: 161752362-161752363 on Build GRCh38
rs777661167 Chr.1: 161752323-161752323 on Build GRCh38
rs757031203 Chr.1: 161752329-161752329 on Build GRCh38
rs748740198 Chr.1: 161752384-161752384 on Build GRCh38
rs999330969 Chr.1: 161752463-161752463 on Build GRCh38
rs576197462 Chr.1: 161752326-161752326 on Build GRCh38
rs1008088506 Chr.1: 161752410-161752410 on Build GRCh38
rs760728897 Chr.1: 161752439-161752439 on Build GRCh38
rs778513093 Chr.1: 161752330-161752330 on Build GRCh38
rs146599576 Chr.1: 161752457-161752457 on Build GRCh38

View Full Product Details