Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31882887-31883132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHMT2
Entrez Gene ID
10919
Gene Name
euchromatic histone lysine methyltransferase 2
Gene Aliases
BAT8, C6orf30, G9A, GAT8, KMT1C, NG36
UniGene
Hs.709218
Gene Chromosome Location
Chr.6: 31879759-31897707 on Build GRCh38
Transcripts
NM_001289413.1 NM_001318833.1 NM_006709.4 NM_025256.6 XM_005248824.3 XM_006714974.1 XM_006714975.1 XM_006714976.1 XM_017010202.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3347131 c.3035C>T p.P1012L Chr.6: 31882969-31882969 on Build GRCh38
4877993 c.3225C>A p.N1075K Chr.6: 31882950-31882950 on Build GRCh38
1131919 c.3054C>A p.N1018K Chr.6: 31882950-31882950 on Build GRCh38
4680501 c.3002G>A p.R1001Q Chr.6: 31883002-31883002 on Build GRCh38
6403558 c.3242G>A p.W1081* Chr.6: 31882933-31882933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771797896 Chr.6: 31882916-31882916 on Build GRCh38
rs759972911 Chr.6: 31882957-31882957 on Build GRCh38
rs907163650 Chr.6: 31883043-31883043 on Build GRCh38
rs957326411 Chr.6: 31883103-31883103 on Build GRCh38
rs770354149 Chr.6: 31883052-31883052 on Build GRCh38
rs562121877 Chr.6: 31883107-31883107 on Build GRCh38
rs749184976 Chr.6: 31883024-31883024 on Build GRCh38
rs201630944 Chr.6: 31882910-31882910 on Build GRCh38
rs776152694 Chr.6: 31883053-31883053 on Build GRCh38
rs757339226 Chr.6: 31883018-31883018 on Build GRCh38
rs778870885 Chr.6: 31883031-31883031 on Build GRCh38
rs751783109 Chr.6: 31883014-31883014 on Build GRCh38
rs758650661 Chr.6: 31882968-31882968 on Build GRCh38
rs760156327 Chr.6: 31882944-31882944 on Build GRCh38
rs980650323 Chr.6: 31883028-31883028 on Build GRCh38
rs372847061 Chr.6: 31883045-31883045 on Build GRCh38
rs115272033 Chr.6: 31883030-31883030 on Build GRCh38
rs142755148 Chr.6: 31882911-31882911 on Build GRCh38
rs547060062 Chr.6: 31883093-31883093 on Build GRCh38
rs3173328 Chr.6: 31882947-31882947 on Build GRCh38
rs147391445 Chr.6: 31882962-31882962 on Build GRCh38
rs1024639968 Chr.6: 31883013-31883013 on Build GRCh38
rs765600287 Chr.6: 31882959-31882959 on Build GRCh38
rs746673755 Chr.6: 31883049-31883049 on Build GRCh38
rs931433235 Chr.6: 31883007-31883007 on Build GRCh38
rs781326113 Chr.6: 31883019-31883019 on Build GRCh38
rs62395847 Chr.6: 31883036-31883036 on Build GRCh38
rs374553342 Chr.6: 31883002-31883002 on Build GRCh38
rs369804736 Chr.6: 31882946-31882946 on Build GRCh38
rs946147081 Chr.6: 31883109-31883109 on Build GRCh38
rs913337498 Chr.6: 31883040-31883040 on Build GRCh38
rs200914483 Chr.6: 31882927-31882927 on Build GRCh38
rs772806733 Chr.6: 31882939-31882939 on Build GRCh38
rs772801707 Chr.6: 31883047-31883047 on Build GRCh38
rs764263583 Chr.6: 31883004-31883004 on Build GRCh38

View Full Product Details