Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 53382065-53382328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIP1L1
Entrez Gene ID
81608
Gene Name
factor interacting with PAPOLA and CPSF1
Gene Aliases
FIP1, Rhe, hFip1
UniGene
Hs.555109
Gene Chromosome Location
Chr.4: 53377572-53462611 on Build GRCh38
Transcripts
NM_001134937.1 NM_001134938.1 NM_030917.3 XM_005265769.4 XM_005265773.4 XM_005265774.4 XM_005265778.4 XM_005265779.4 XM_005265781.4 XM_005265782.4 XM_017008662.1 XM_017008663.1 XM_017008664.1 XM_017008665.1 XM_017008666.1 XM_017008667.1 XM_017008668.1 XM_017008669.1 XM_017008670.1 XM_017008671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4802639 c.154G>C p.E52Q Chr.4: 53382306-53382306 on Build GRCh38
4802638 c.199G>C p.E67Q Chr.4: 53382306-53382306 on Build GRCh38
6445030 c.217G>A p.V73I Chr.4: 53382324-53382324 on Build GRCh38
734195 c.215G>T p.G72V Chr.4: 53382322-53382322 on Build GRCh38
6836335 c.136C>G p.P46A Chr.4: 53382288-53382288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897361567 Chr.4: 53382130-53382130 on Build GRCh38
rs1021724667 Chr.4: 53382285-53382285 on Build GRCh38
rs1011925673 Chr.4: 53382193-53382193 on Build GRCh38
rs759704712 Chr.4: 53382263-53382263 on Build GRCh38
rs377691648 Chr.4: 53382244-53382244 on Build GRCh38
rs746635224 Chr.4: 53382129-53382129 on Build GRCh38
rs985836459 Chr.4: 53382098-53382098 on Build GRCh38
rs758179094 Chr.4: 53382215-53382215 on Build GRCh38
rs1041278419 Chr.4: 53382115-53382115 on Build GRCh38
rs775635500 Chr.4: 53382274-53382274 on Build GRCh38
rs777298870 Chr.4: 53382110-53382110 on Build GRCh38
rs374562663 Chr.4: 53382237-53382237 on Build GRCh38
rs997832 Chr.4: 53382109-53382109 on Build GRCh38
rs369457633 Chr.4: 53382234-53382234 on Build GRCh38
rs746072301 Chr.4: 53382247-53382247 on Build GRCh38
rs113304970 Chr.4: 53382233-53382233 on Build GRCh38
rs747321414 Chr.4: 53382238-53382238 on Build GRCh38
rs1023686965 Chr.4: 53382200-53382200 on Build GRCh38
rs774419463 Chr.4: 53382256-53382259 on Build GRCh38
rs930132530 Chr.4: 53382139-53382139 on Build GRCh38
rs566644126 Chr.4: 53382146-53382147 on Build GRCh38
rs879030585 Chr.4: 53382265-53382265 on Build GRCh38
rs56007140 Chr.4: 53382251-53382251 on Build GRCh38
rs570690766 Chr.4: 53382186-53382186 on Build GRCh38
rs112825339 Chr.4: 53382144-53382144 on Build GRCh38
rs183271016 Chr.4: 53382239-53382239 on Build GRCh38
rs888167037 Chr.4: 53382188-53382188 on Build GRCh38
rs764777198 Chr.4: 53382228-53382228 on Build GRCh38
rs911486548 Chr.4: 53382101-53382101 on Build GRCh38
rs748082351 Chr.4: 53382288-53382288 on Build GRCh38

View Full Product Details