Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153951253-153951526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC2
Entrez Gene ID
200186
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
TORC-2, TORC2
UniGene
Hs.406392
Gene Chromosome Location
Chr.1: 153947672-153958656 on Build GRCh38
Transcripts
NM_181715.2 XM_005244946.1 XM_005244949.2 XM_017000575.1 XM_017000576.1 XM_017000577.1 XM_017000578.1 XM_017000579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6423674 c.1140T>A p.H380Q Chr.1: 153951524-153951524 on Build GRCh38
4598445 c.1226C>T p.P409L Chr.1: 153951438-153951438 on Build GRCh38
3474558 c.1155C>T p.T385T Chr.1: 153951509-153951509 on Build GRCh38
1334349 c.1282C>T p.R428C Chr.1: 153951382-153951382 on Build GRCh38
5461997 c.1397T>C p.I466T Chr.1: 153951267-153951267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759201075 Chr.1: 153951443-153951443 on Build GRCh38
rs890035726 Chr.1: 153951427-153951427 on Build GRCh38
rs145272706 Chr.1: 153951338-153951338 on Build GRCh38
rs201467539 Chr.1: 153951425-153951425 on Build GRCh38
rs754577717 Chr.1: 153951297-153951297 on Build GRCh38
rs368360023 Chr.1: 153951448-153951448 on Build GRCh38
rs773243105 Chr.1: 153951419-153951419 on Build GRCh38
rs563056498 Chr.1: 153951296-153951296 on Build GRCh38
rs762839321 Chr.1: 153951368-153951368 on Build GRCh38
rs957020470 Chr.1: 153951487-153951487 on Build GRCh38
rs781759272 Chr.1: 153951311-153951311 on Build GRCh38
rs1049972247 Chr.1: 153951420-153951420 on Build GRCh38
rs757600510 Chr.1: 153951470-153951470 on Build GRCh38
rs902662579 Chr.1: 153951441-153951441 on Build GRCh38
rs377025190 Chr.1: 153951378-153951378 on Build GRCh38
rs776532141 Chr.1: 153951352-153951352 on Build GRCh38
rs746142911 Chr.1: 153951405-153951405 on Build GRCh38
rs774234159 Chr.1: 153951508-153951508 on Build GRCh38
rs749306326 Chr.1: 153951506-153951506 on Build GRCh38
rs17854826 Chr.1: 153951473-153951473 on Build GRCh38
rs752259202 Chr.1: 153951388-153951388 on Build GRCh38
rs780041213 Chr.1: 153951408-153951408 on Build GRCh38
rs895249711 Chr.1: 153951467-153951467 on Build GRCh38
rs764473936 Chr.1: 153951456-153951456 on Build GRCh38
rs202092600 Chr.1: 153951396-153951396 on Build GRCh38
rs758755660 Chr.1: 153951387-153951388 on Build GRCh38
rs769773917 Chr.1: 153951363-153951363 on Build GRCh38
rs201148360 Chr.1: 153951428-153951428 on Build GRCh38
rs574064798 Chr.1: 153951379-153951379 on Build GRCh38
rs759103873 Chr.1: 153951380-153951380 on Build GRCh38
rs774143497 Chr.1: 153951429-153951429 on Build GRCh38
rs149147178 Chr.1: 153951350-153951350 on Build GRCh38
rs765653312 Chr.1: 153951476-153951476 on Build GRCh38
rs761723625 Chr.1: 153951362-153951362 on Build GRCh38
rs767294894 Chr.1: 153951447-153951447 on Build GRCh38
rs752337255 Chr.1: 153951450-153951461 on Build GRCh38
rs372938693 Chr.1: 153951389-153951389 on Build GRCh38
rs771810106 Chr.1: 153951499-153951499 on Build GRCh38
rs766791516 Chr.1: 153951449-153951451 on Build GRCh38
rs777410003 Chr.1: 153951462-153951464 on Build GRCh38
rs754415307 Chr.1: 153951458-153951458 on Build GRCh38
rs772133880 Chr.1: 153951406-153951406 on Build GRCh38
rs757655663 Chr.1: 153951399-153951399 on Build GRCh38
rs772823934 Chr.1: 153951364-153951364 on Build GRCh38
rs139505722 Chr.1: 153951334-153951334 on Build GRCh38
rs747698969 Chr.1: 153951349-153951349 on Build GRCh38
rs566531709 Chr.1: 153951496-153951496 on Build GRCh38
rs142680858 Chr.1: 153951406-153951413 on Build GRCh38
rs1042624658 Chr.1: 153951439-153951439 on Build GRCh38
rs780343779 Chr.1: 153951486-153951486 on Build GRCh38
rs779081744 Chr.1: 153951403-153951403 on Build GRCh38
rs750790359 Chr.1: 153951479-153951479 on Build GRCh38
rs765723874 Chr.1: 153951288-153951290 on Build GRCh38
rs747030449 Chr.1: 153951492-153951492 on Build GRCh38
rs753202757 Chr.1: 153951321-153951321 on Build GRCh38
rs766140950 Chr.1: 153951283-153951283 on Build GRCh38
rs769567701 Chr.1: 153951415-153951415 on Build GRCh38
rs200739285 Chr.1: 153951507-153951507 on Build GRCh38
rs749680802 Chr.1: 153951337-153951337 on Build GRCh38
rs149725052 Chr.1: 153951335-153951335 on Build GRCh38
rs372705085 Chr.1: 153951455-153951455 on Build GRCh38
rs750938869 Chr.1: 153951303-153951306 on Build GRCh38
rs756523382 Chr.1: 153951391-153951391 on Build GRCh38
rs778369404 Chr.1: 153951395-153951395 on Build GRCh38
rs758557646 Chr.1: 153951480-153951480 on Build GRCh38
rs568652768 Chr.1: 153951381-153951381 on Build GRCh38
rs755709644 Chr.1: 153951449-153951450 on Build GRCh38
rs779135391 Chr.1: 153951348-153951348 on Build GRCh38
rs1037455921 Chr.1: 153951343-153951343 on Build GRCh38
rs1008385072 Chr.1: 153951448-153951449 on Build GRCh38
rs975832658 Chr.1: 153951347-153951347 on Build GRCh38

View Full Product Details