Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44219917-44220160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMAP1
Entrez Gene ID
55929
Gene Name
DNA methyltransferase 1 associated protein 1
Gene Aliases
DNMAP1, DNMTAP1, EAF2, MEAF2, SWC4
UniGene
Hs.8008
Gene Chromosome Location
Chr.1: 44213428-44220679 on Build GRCh38
Transcripts
NM_001034023.1 NM_001034024.1 NM_019100.4 XM_005271037.4 XM_005271039.1 XM_005271040.1 XM_006710771.3 XM_011541784.2 XM_011541785.2 XM_017001807.1 XM_017001808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
910001 c.1175G>A p.R392Q Chr.1: 44220140-44220140 on Build GRCh38
4008367 c.1070C>T p.T357M Chr.1: 44220035-44220035 on Build GRCh38
4978028 c.1180C>T p.R394C Chr.1: 44220145-44220145 on Build GRCh38
1342961 c.1103G>A p.R368Q Chr.1: 44220068-44220068 on Build GRCh38
4008368 c.1107C>T p.S369S Chr.1: 44220072-44220072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146514924 Chr.1: 44220138-44220138 on Build GRCh38
rs766235875 Chr.1: 44219967-44219967 on Build GRCh38
rs763717827 Chr.1: 44220118-44220118 on Build GRCh38
rs41270415 Chr.1: 44219992-44219992 on Build GRCh38
rs773525336 Chr.1: 44220002-44220002 on Build GRCh38
rs746310575 Chr.1: 44219995-44219995 on Build GRCh38
rs777229352 Chr.1: 44220079-44220079 on Build GRCh38
rs752229758 Chr.1: 44220072-44220072 on Build GRCh38
rs767182825 Chr.1: 44220068-44220068 on Build GRCh38
rs748252842 Chr.1: 44220123-44220123 on Build GRCh38
rs368180564 Chr.1: 44219991-44219991 on Build GRCh38
rs758418548 Chr.1: 44220010-44220010 on Build GRCh38
rs569718563 Chr.1: 44220048-44220048 on Build GRCh38
rs117340264 Chr.1: 44220096-44220096 on Build GRCh38
rs564972388 Chr.1: 44219979-44219979 on Build GRCh38
rs189650822 Chr.1: 44220075-44220075 on Build GRCh38
rs116432610 Chr.1: 44219971-44219971 on Build GRCh38
rs878922277 Chr.1: 44219941-44219941 on Build GRCh38
rs116305257 Chr.1: 44220053-44220053 on Build GRCh38
rs759314123 Chr.1: 44220054-44220054 on Build GRCh38
rs768962026 Chr.1: 44220098-44220098 on Build GRCh38
rs951792544 Chr.1: 44220120-44220120 on Build GRCh38
rs879241457 Chr.1: 44219951-44219951 on Build GRCh38
rs999952777 Chr.1: 44220037-44220037 on Build GRCh38
rs777274603 Chr.1: 44219994-44219994 on Build GRCh38
rs769937212 Chr.1: 44220029-44220029 on Build GRCh38
rs74983620 Chr.1: 44219989-44219989 on Build GRCh38
rs116279448 Chr.1: 44220090-44220090 on Build GRCh38
rs147610607 Chr.1: 44220005-44220005 on Build GRCh38
rs773455439 Chr.1: 44220036-44220036 on Build GRCh38
rs917343309 Chr.1: 44220009-44220009 on Build GRCh38
rs149066039 Chr.1: 44220024-44220024 on Build GRCh38
rs762934376 Chr.1: 44220043-44220043 on Build GRCh38
rs965373926 Chr.1: 44219953-44219953 on Build GRCh38
rs753373766 Chr.1: 44220087-44220087 on Build GRCh38
rs368073383 Chr.1: 44220001-44220001 on Build GRCh38
rs752244753 Chr.1: 44219993-44219993 on Build GRCh38
rs761939880 Chr.1: 44220027-44220027 on Build GRCh38
rs202183088 Chr.1: 44219988-44219988 on Build GRCh38
rs997898480 Chr.1: 44219959-44219959 on Build GRCh38
rs1015359608 Chr.1: 44219938-44219938 on Build GRCh38
rs781630711 Chr.1: 44220102-44220102 on Build GRCh38
rs138593911 Chr.1: 44220023-44220023 on Build GRCh38

View Full Product Details