Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49365452-49365723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKKL1
Entrez Gene ID
27120
Gene Name
dickkopf like acrosomal protein 1
Gene Aliases
CT34, SGY, SGY-1, SGY1
UniGene
Hs.515855
Gene Chromosome Location
Chr.19: 49360520-49375132 on Build GRCh38
Transcripts
NM_001197301.1 NM_001197302.1 NM_014419.3 XM_006723142.2 XM_011526724.1 XM_011526725.1 XM_011526726.2 XM_011526727.2 XM_011526728.2 XM_011526729.1 XM_017026593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7147025 c.196C>T p.R66W Chr.19: 49365521-49365521 on Build GRCh38
1250138 c.319G>A p.D107N Chr.19: 49365644-49365644 on Build GRCh38
1325138 c.287G>T p.G96V Chr.19: 49365612-49365612 on Build GRCh38
4080043 c.295A>G p.T99A Chr.19: 49365620-49365620 on Build GRCh38
5687652 c.274G>T p.E92* Chr.19: 49365599-49365599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769308683 Chr.19: 49365514-49365514 on Build GRCh38
rs774183266 Chr.19: 49365624-49365624 on Build GRCh38
rs781174561 Chr.19: 49365545-49365545 on Build GRCh38
rs148116480 Chr.19: 49365561-49365561 on Build GRCh38
rs766416135 Chr.19: 49365653-49365653 on Build GRCh38
rs748501492 Chr.19: 49365590-49365590 on Build GRCh38
rs748104338 Chr.19: 49365557-49365557 on Build GRCh38
rs201665269 Chr.19: 49365494-49365494 on Build GRCh38
rs768963206 Chr.19: 49365500-49365500 on Build GRCh38
rs759271501 Chr.19: 49365526-49365526 on Build GRCh38
rs747732578 Chr.19: 49365508-49365508 on Build GRCh38
rs753097782 Chr.19: 49365678-49365678 on Build GRCh38
rs759448619 Chr.19: 49365632-49365632 on Build GRCh38
rs781663426 Chr.19: 49365674-49365674 on Build GRCh38
rs748798280 Chr.19: 49365607-49365607 on Build GRCh38
rs776298251 Chr.19: 49365501-49365501 on Build GRCh38
rs779066964 Chr.19: 49365480-49365480 on Build GRCh38
rs750512153 Chr.19: 49365560-49365560 on Build GRCh38
rs2303758 Chr.19: 49365702-49365702 on Build GRCh38
rs967848672 Chr.19: 49365482-49365482 on Build GRCh38
rs755538208 Chr.19: 49365668-49365668 on Build GRCh38
rs752021470 Chr.19: 49365667-49365667 on Build GRCh38
rs920565008 Chr.19: 49365666-49365666 on Build GRCh38
rs746397580 Chr.19: 49365489-49365489 on Build GRCh38
rs747386126 Chr.19: 49365497-49365497 on Build GRCh38
rs749502791 Chr.19: 49365619-49365619 on Build GRCh38
rs777311923 Chr.19: 49365696-49365696 on Build GRCh38
rs759727217 Chr.19: 49365680-49365680 on Build GRCh38
rs537755411 Chr.19: 49365542-49365542 on Build GRCh38
rs894246649 Chr.19: 49365602-49365602 on Build GRCh38
rs772768508 Chr.19: 49365517-49365517 on Build GRCh38
rs35693137 Chr.19: 49365635-49365635 on Build GRCh38
rs747514090 Chr.19: 49365575-49365575 on Build GRCh38
rs141709326 Chr.19: 49365580-49365580 on Build GRCh38
rs746574001 Chr.19: 49365490-49365490 on Build GRCh38
rs146176366 Chr.19: 49365644-49365644 on Build GRCh38
rs986238125 Chr.19: 49365701-49365701 on Build GRCh38
rs767431412 Chr.19: 49365633-49365633 on Build GRCh38
rs762979135 Chr.19: 49365652-49365652 on Build GRCh38
rs34625472 Chr.19: 49365643-49365643 on Build GRCh38
rs975413217 Chr.19: 49365532-49365532 on Build GRCh38
rs772888564 Chr.19: 49365601-49365601 on Build GRCh38
rs773505700 Chr.19: 49365514-49365514 on Build GRCh38
rs748930330 Chr.19: 49365698-49365698 on Build GRCh38
rs767376449 Chr.19: 49365528-49365528 on Build GRCh38
rs757494010 Chr.19: 49365530-49365530 on Build GRCh38
rs757439165 Chr.19: 49365476-49365476 on Build GRCh38
rs780451353 Chr.19: 49365565-49365565 on Build GRCh38
rs150574340 Chr.19: 49365612-49365612 on Build GRCh38
rs756446313 Chr.19: 49365689-49365689 on Build GRCh38

View Full Product Details