Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15231082-15231307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX3
Entrez Gene ID
79852
Gene Name
epoxide hydrolase 3
Gene Aliases
ABHD9, EH3
UniGene
Hs.156457
Gene Chromosome Location
Chr.19: 15226919-15233047 on Build GRCh38
Transcripts
NM_001142886.1 NM_024794.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992118 c.494C>T p.S165L Chr.19: 15231084-15231084 on Build GRCh38
6043235 c.487G>A p.G163S Chr.19: 15231239-15231239 on Build GRCh38
1480703 c.445G>C p.D149H Chr.19: 15231281-15231281 on Build GRCh38
6017333 c.481G>A p.G161S Chr.19: 15231245-15231245 on Build GRCh38
4496554 c.477C>T p.I159I Chr.19: 15231249-15231249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745616505 Chr.19: 15231240-15231240 on Build GRCh38
rs764767555 Chr.19: 15231213-15231213 on Build GRCh38
rs181026961 Chr.19: 15231282-15231282 on Build GRCh38
rs952951925 Chr.19: 15231148-15231148 on Build GRCh38
rs560863212 Chr.19: 15231182-15231182 on Build GRCh38
rs762041019 Chr.19: 15231266-15231266 on Build GRCh38
rs752444762 Chr.19: 15231225-15231225 on Build GRCh38
rs769575823 Chr.19: 15231232-15231232 on Build GRCh38
rs201984223 Chr.19: 15231111-15231111 on Build GRCh38
rs933702302 Chr.19: 15231145-15231145 on Build GRCh38
rs780086523 Chr.19: 15231250-15231250 on Build GRCh38
rs769676805 Chr.19: 15231244-15231244 on Build GRCh38
rs1050787218 Chr.19: 15231161-15231161 on Build GRCh38
rs757797722 Chr.19: 15231230-15231230 on Build GRCh38
rs772700060 Chr.19: 15231270-15231270 on Build GRCh38
rs780506628 Chr.19: 15231188-15231188 on Build GRCh38
rs908186241 Chr.19: 15231173-15231173 on Build GRCh38
rs762915627 Chr.19: 15231114-15231114 on Build GRCh38
rs768788890 Chr.19: 15231257-15231257 on Build GRCh38
rs749551343 Chr.19: 15231252-15231252 on Build GRCh38
rs761171167 Chr.19: 15231201-15231201 on Build GRCh38
rs774959495 Chr.19: 15231129-15231129 on Build GRCh38
rs777300826 Chr.19: 15231231-15231231 on Build GRCh38
rs757202119 Chr.19: 15231233-15231233 on Build GRCh38
rs373635598 Chr.19: 15231265-15231265 on Build GRCh38
rs764287058 Chr.19: 15231125-15231125 on Build GRCh38
rs775819468 Chr.19: 15231113-15231113 on Build GRCh38
rs922241814 Chr.19: 15231214-15231214 on Build GRCh38
rs540549600 Chr.19: 15231178-15231178 on Build GRCh38
rs776418220 Chr.19: 15231108-15231108 on Build GRCh38
rs147393873 Chr.19: 15231189-15231189 on Build GRCh38
rs781166308 Chr.19: 15231237-15231237 on Build GRCh38

View Full Product Details