Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109606353-109606598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT2
Entrez Gene ID
2780
Gene Name
G protein subunit alpha transducin 2
Gene Aliases
ACHM4, GNATC
UniGene
Hs.36973
Gene Chromosome Location
Chr.1: 109603091-109619733 on Build GRCh38
Transcripts
NM_005272.3 XM_011541264.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1743423 c.485T>G p.I162S Chr.1: 109606413-109606413 on Build GRCh38
6049095 c.472C>G p.Q158E Chr.1: 109606426-109606426 on Build GRCh38
3470939 c.540A>T p.K180N Chr.1: 109606358-109606358 on Build GRCh38
7137034 c.482G>T p.R161L Chr.1: 109606416-109606416 on Build GRCh38
674454 c.525C>G p.L175L Chr.1: 109606373-109606373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878924335 Chr.1: 109606503-109606503 on Build GRCh38
rs377203942 Chr.1: 109606479-109606479 on Build GRCh38
rs528785489 Chr.1: 109606512-109606512 on Build GRCh38
rs766033906 Chr.1: 109606435-109606435 on Build GRCh38
rs759522552 Chr.1: 109606394-109606395 on Build GRCh38
rs948907259 Chr.1: 109606528-109606528 on Build GRCh38
rs1002057655 Chr.1: 109606493-109606493 on Build GRCh38
rs904304609 Chr.1: 109606540-109606540 on Build GRCh38
rs760195431 Chr.1: 109606433-109606433 on Build GRCh38
rs1028161660 Chr.1: 109606390-109606390 on Build GRCh38
rs894720415 Chr.1: 109606516-109606516 on Build GRCh38
rs909323361 Chr.1: 109606424-109606424 on Build GRCh38
rs1043230631 Chr.1: 109606535-109606535 on Build GRCh38
rs763373350 Chr.1: 109606447-109606447 on Build GRCh38
rs747669348 Chr.1: 109606400-109606400 on Build GRCh38
rs147524117 Chr.1: 109606492-109606492 on Build GRCh38
rs775763827 Chr.1: 109606441-109606441 on Build GRCh38
rs768651686 Chr.1: 109606388-109606388 on Build GRCh38
rs983915440 Chr.1: 109606416-109606416 on Build GRCh38
rs752112184 Chr.1: 109606455-109606455 on Build GRCh38
rs183584111 Chr.1: 109606452-109606452 on Build GRCh38
rs1033962155 Chr.1: 109606506-109606506 on Build GRCh38
rs757329539 Chr.1: 109606461-109606461 on Build GRCh38
rs756237086 Chr.1: 109606484-109606484 on Build GRCh38
rs772147864 Chr.1: 109606429-109606429 on Build GRCh38
rs774485451 Chr.1: 109606398-109606398 on Build GRCh38
rs942106145 Chr.1: 109606427-109606427 on Build GRCh38
rs953848102 Chr.1: 109606405-109606405 on Build GRCh38
rs767457006 Chr.1: 109606474-109606474 on Build GRCh38
rs745308973 Chr.1: 109606417-109606417 on Build GRCh38
rs771518962 Chr.1: 109606403-109606403 on Build GRCh38

View Full Product Details