Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30893676-30893905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL7C
Entrez Gene ID
9274
Gene Name
BCL tumor suppressor 7C
Gene Aliases
-
UniGene
Hs.658547
Gene Chromosome Location
Chr.16: 30834041-30894550 on Build GRCh38
Transcripts
NM_001286526.1 NM_004765.3 XM_011545980.2 XM_017023885.1 XM_017023886.1 XM_017023887.1 XM_017023888.1 NR_031576.1 NR_110940.1 NR_110941.1 NR_110942.1

Gene Symbol
MIR762
Entrez Gene ID
100313837
Gene Name
microRNA 762
Gene Aliases
hsa-mir-762
UniGene
Gene Chromosome Location
Chr.16: 30893903-30893985 on Build GRCh38
Transcripts
NM_001286526.1 NM_004765.3 XM_011545980.2 XM_017023885.1 XM_017023886.1 XM_017023887.1 XM_017023888.1 NR_031576.1 NR_110940.1 NR_110941.1 NR_110942.1

Gene Symbol
MIR762HG
Entrez Gene ID
101928736
Gene Name
MIR762 host gene
Gene Aliases
-
UniGene
Hs.303197
Gene Chromosome Location
Chr.16: 30875460-30895220 on Build GRCh38
Transcripts
NM_001286526.1 NM_004765.3 XM_011545980.2 XM_017023885.1 XM_017023886.1 XM_017023887.1 XM_017023888.1 NR_031576.1 NR_110940.1 NR_110941.1 NR_110942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6378309 c.47A>G p.D16G Chr.16: 30893898-30893898 on Build GRCh38
5054848 c.84C>A p.V28V Chr.16: 30893861-30893861 on Build GRCh38
6191147 c.71delC p.I25fs*12 Chr.16: 30893874-30893874 on Build GRCh38
5701396 c.71_72insA p.I25fs*69 Chr.16: 30893873-30893874 on Build GRCh38
5687506 c.71C>A p.T24N Chr.16: 30893874-30893874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017480170 Chr.16: 30893741-30893741 on Build GRCh38
rs3794707 Chr.16: 30893726-30893726 on Build GRCh38
rs759527404 Chr.16: 30893827-30893827 on Build GRCh38
rs924360181 Chr.16: 30893784-30893784 on Build GRCh38
rs764469929 Chr.16: 30893814-30893814 on Build GRCh38
rs747017961 Chr.16: 30893868-30893868 on Build GRCh38
rs750888197 Chr.16: 30893842-30893842 on Build GRCh38
rs978509642 Chr.16: 30893762-30893762 on Build GRCh38
rs200155793 Chr.16: 30893878-30893878 on Build GRCh38
rs1007477945 Chr.16: 30893711-30893711 on Build GRCh38
rs1039780723 Chr.16: 30893707-30893707 on Build GRCh38
rs754376558 Chr.16: 30893832-30893832 on Build GRCh38
rs925324830 Chr.16: 30893756-30893756 on Build GRCh38
rs758090967 Chr.16: 30893839-30893839 on Build GRCh38
rs955910624 Chr.16: 30893786-30893786 on Build GRCh38
rs757641673 Chr.16: 30893834-30893834 on Build GRCh38
rs936686943 Chr.16: 30893768-30893768 on Build GRCh38
rs780442592 Chr.16: 30893852-30893852 on Build GRCh38
rs968505794 Chr.16: 30893759-30893759 on Build GRCh38
rs768783625 Chr.16: 30893872-30893872 on Build GRCh38
rs758667437 Chr.16: 30893846-30893846 on Build GRCh38
rs892395553 Chr.16: 30893783-30893783 on Build GRCh38
rs1052362011 Chr.16: 30893772-30893772 on Build GRCh38
rs545588513 Chr.16: 30893881-30893881 on Build GRCh38
rs571935222 Chr.16: 30893700-30893700 on Build GRCh38
rs759109113 Chr.16: 30893804-30893804 on Build GRCh38

View Full Product Details