Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 122981311-122981558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSX
Entrez Gene ID
390259
Gene Name
brain specific homeobox
Gene Aliases
BSX1
UniGene
Hs.449687
Gene Chromosome Location
Chr.11: 122977649-122981671 on Build GRCh38
Transcripts
NM_001098169.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3720991 c.185C>A p.P62H Chr.11: 122981487-122981487 on Build GRCh38
6425987 c.260C>T p.S87L Chr.11: 122981412-122981412 on Build GRCh38
924143 c.260C>A p.S87* Chr.11: 122981412-122981412 on Build GRCh38
924144 c.239A>C p.H80P Chr.11: 122981433-122981433 on Build GRCh38
5589847 c.238C>T p.H80Y Chr.11: 122981434-122981434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199643758 Chr.11: 122981476-122981476 on Build GRCh38
rs1012698643 Chr.11: 122981466-122981466 on Build GRCh38
rs868246509 Chr.11: 122981412-122981412 on Build GRCh38
rs771464195 Chr.11: 122981495-122981495 on Build GRCh38
rs749798967 Chr.11: 122981487-122981487 on Build GRCh38
rs751359449 Chr.11: 122981400-122981400 on Build GRCh38
rs971109678 Chr.11: 122981502-122981502 on Build GRCh38
rs1022741036 Chr.11: 122981492-122981492 on Build GRCh38
rs543949010 Chr.11: 122981395-122981395 on Build GRCh38
rs372663917 Chr.11: 122981468-122981468 on Build GRCh38
rs200602573 Chr.11: 122981372-122981372 on Build GRCh38
rs775462840 Chr.11: 122981387-122981388 on Build GRCh38
rs1054734813 Chr.11: 122981430-122981430 on Build GRCh38
rs762860042 Chr.11: 122981525-122981525 on Build GRCh38
rs746254703 Chr.11: 122981500-122981500 on Build GRCh38
rs200026871 Chr.11: 122981504-122981504 on Build GRCh38
rs760509770 Chr.11: 122981470-122981472 on Build GRCh38
rs780996854 Chr.11: 122981440-122981440 on Build GRCh38
rs867300572 Chr.11: 122981529-122981529 on Build GRCh38
rs774634223 Chr.11: 122981499-122981499 on Build GRCh38
rs760833595 Chr.11: 122981509-122981509 on Build GRCh38
rs553936426 Chr.11: 122981337-122981337 on Build GRCh38
rs772352727 Chr.11: 122981503-122981503 on Build GRCh38
rs370019336 Chr.11: 122981411-122981411 on Build GRCh38
rs779783062 Chr.11: 122981396-122981396 on Build GRCh38
rs934011983 Chr.11: 122981335-122981335 on Build GRCh38
rs763767296 Chr.11: 122981459-122981459 on Build GRCh38
rs867242555 Chr.11: 122981475-122981475 on Build GRCh38
rs975383627 Chr.11: 122981346-122981346 on Build GRCh38
rs892963804 Chr.11: 122981444-122981444 on Build GRCh38
rs913951255 Chr.11: 122981443-122981443 on Build GRCh38
rs370532557 Chr.11: 122981516-122981516 on Build GRCh38
rs776770866 Chr.11: 122981524-122981524 on Build GRCh38
rs764114824 Chr.11: 122981368-122981368 on Build GRCh38

View Full Product Details