Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11454371-11454602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELAVL3
Entrez Gene ID
1995
Gene Name
ELAV like neuron-specific RNA binding protein 3
Gene Aliases
HUC, HUCL, PLE21
UniGene
Hs.1701
Gene Chromosome Location
Chr.19: 11451328-11480994 on Build GRCh38
Transcripts
NM_001420.3 NM_032281.2 XM_011527778.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5013372 c.1060G>A p.V354M Chr.19: 11454570-11454570 on Build GRCh38
3388579 c.1059C>T p.R353R Chr.19: 11454571-11454571 on Build GRCh38
5780458 c.1045C>T p.R349C Chr.19: 11454585-11454585 on Build GRCh38
6653167 c.1039G>A p.G347S Chr.19: 11454591-11454591 on Build GRCh38
110441 c.1074C>T p.S358S Chr.19: 11454556-11454556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778492575 Chr.19: 11454530-11454530 on Build GRCh38
rs966855065 Chr.19: 11454450-11454451 on Build GRCh38
rs911450726 Chr.19: 11454469-11454470 on Build GRCh38
rs757745976 Chr.19: 11454567-11454567 on Build GRCh38
rs1016073271 Chr.19: 11454437-11454437 on Build GRCh38
rs145408161 Chr.19: 11454576-11454576 on Build GRCh38
rs926361415 Chr.19: 11454577-11454577 on Build GRCh38
rs779645608 Chr.19: 11454523-11454523 on Build GRCh38
rs760668510 Chr.19: 11454499-11454499 on Build GRCh38
rs747153913 Chr.19: 11454583-11454583 on Build GRCh38
rs936882896 Chr.19: 11454452-11454469 on Build GRCh38
rs757797512 Chr.19: 11454521-11454521 on Build GRCh38
rs748089092 Chr.19: 11454472-11454473 on Build GRCh38
rs963554329 Chr.19: 11454467-11454467 on Build GRCh38
rs925424662 Chr.19: 11454450-11454457 on Build GRCh38
rs369778910 Chr.19: 11454399-11454399 on Build GRCh38
rs191710732 Chr.19: 11454571-11454571 on Build GRCh38
rs764834247 Chr.19: 11454509-11454509 on Build GRCh38
rs35334288 Chr.19: 11454470-11454471 on Build GRCh38
rs977908007 Chr.19: 11454450-11454453 on Build GRCh38
rs767566028 Chr.19: 11454470-11454473 on Build GRCh38
rs568484090 Chr.19: 11454493-11454493 on Build GRCh38
rs1024655825 Chr.19: 11454449-11454449 on Build GRCh38
rs145003649 Chr.19: 11454556-11454556 on Build GRCh38
rs991785064 Chr.19: 11454435-11454435 on Build GRCh38
rs775255481 Chr.19: 11454497-11454497 on Build GRCh38
rs1007789478 Chr.19: 11454416-11454416 on Build GRCh38
rs139974609 Chr.19: 11454529-11454529 on Build GRCh38
rs901135573 Chr.19: 11454396-11454396 on Build GRCh38
rs753613748 Chr.19: 11454501-11454501 on Build GRCh38
rs547037629 Chr.19: 11454412-11454412 on Build GRCh38
rs749641214 Chr.19: 11454547-11454547 on Build GRCh38
rs761661246 Chr.19: 11454502-11454502 on Build GRCh38
rs200383181 Chr.19: 11454500-11454500 on Build GRCh38
rs985605661 Chr.19: 11454468-11454469 on Build GRCh38
rs147698039 Chr.19: 11454580-11454580 on Build GRCh38
rs753076296 Chr.19: 11454527-11454527 on Build GRCh38
rs778955437 Chr.19: 11454570-11454570 on Build GRCh38
rs750136144 Chr.19: 11454517-11454517 on Build GRCh38
rs1049833660 Chr.19: 11454408-11454408 on Build GRCh38

View Full Product Details