Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 24505099-24505330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH10
Entrez Gene ID
1008
Gene Name
cadherin 10
Gene Aliases
-
UniGene
Hs.92489
Gene Chromosome Location
Chr.5: 24487100-24644978 on Build GRCh38
Transcripts
NM_001317222.1 NM_001317224.1 NM_006727.4 XM_011513923.2 XM_017008915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5647406 c.1257-1G>C p.? Chr.5: 24505249-24505249 on Build GRCh38
387532 c.1283T>A p.L428H Chr.5: 24505222-24505222 on Build GRCh38
1311081 c.1293C>G p.I431M Chr.5: 24505212-24505212 on Build GRCh38
6624583 c.1316G>A p.G439E Chr.5: 24505189-24505189 on Build GRCh38
4997650 c.1283T>G p.L428R Chr.5: 24505222-24505222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866850339 Chr.5: 24505202-24505202 on Build GRCh38
rs903128657 Chr.5: 24505223-24505223 on Build GRCh38
rs745564475 Chr.5: 24505185-24505185 on Build GRCh38
rs745687203 Chr.5: 24505296-24505296 on Build GRCh38
rs780952857 Chr.5: 24505266-24505267 on Build GRCh38
rs375518042 Chr.5: 24505151-24505151 on Build GRCh38
rs376630462 Chr.5: 24505248-24505248 on Build GRCh38
rs760235178 Chr.5: 24505214-24505214 on Build GRCh38
rs149199529 Chr.5: 24505152-24505152 on Build GRCh38
rs376724275 Chr.5: 24505289-24505289 on Build GRCh38
rs770913066 Chr.5: 24505168-24505168 on Build GRCh38
rs555555015 Chr.5: 24505241-24505241 on Build GRCh38
rs752659067 Chr.5: 24505276-24505276 on Build GRCh38
rs544116954 Chr.5: 24505146-24505146 on Build GRCh38
rs566574417 Chr.5: 24505297-24505297 on Build GRCh38
rs551532614 Chr.5: 24505283-24505283 on Build GRCh38
rs144680689 Chr.5: 24505262-24505262 on Build GRCh38
rs755023658 Chr.5: 24505133-24505133 on Build GRCh38
rs372226944 Chr.5: 24505208-24505208 on Build GRCh38
rs771873502 Chr.5: 24505196-24505196 on Build GRCh38
rs111297824 Chr.5: 24505292-24505292 on Build GRCh38
rs770143615 Chr.5: 24505234-24505234 on Build GRCh38
rs202215608 Chr.5: 24505298-24505298 on Build GRCh38
rs767814386 Chr.5: 24505273-24505273 on Build GRCh38
rs756273162 Chr.5: 24505282-24505282 on Build GRCh38
rs746488894 Chr.5: 24505299-24505299 on Build GRCh38
rs373822384 Chr.5: 24505288-24505288 on Build GRCh38
rs894221809 Chr.5: 24505130-24505130 on Build GRCh38
rs1042728194 Chr.5: 24505180-24505180 on Build GRCh38
rs763303674 Chr.5: 24505258-24505258 on Build GRCh38
rs749021068 Chr.5: 24505159-24505159 on Build GRCh38
rs777777744 Chr.5: 24505158-24505158 on Build GRCh38
rs759709217 Chr.5: 24505269-24505269 on Build GRCh38
rs778672806 Chr.5: 24505169-24505169 on Build GRCh38

View Full Product Details