Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44155237-44155474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
UniGene
Hs.437059
Gene Chromosome Location
Chr.17: 44141902-44162477 on Build GRCh38
Transcripts
NM_001171251.2 NM_001321310.1 NM_001321311.1 NM_024032.4 XM_011525187.1 XM_011525188.1 XM_011525189.1 XM_011525190.1 XM_011525191.2 XM_011525192.1 XM_011525193.1 XM_011525195.1 XM_011525197.2 XM_017025031.1 XM_017025032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1679860 c.1688A>G p.N563S Chr.17: 44155326-44155326 on Build GRCh38
979889 c.1672C>T p.R558* Chr.17: 44155310-44155310 on Build GRCh38
1198237 c.1673G>A p.R558Q Chr.17: 44155311-44155311 on Build GRCh38
1684509 c.1656_1657delGT p.P555fs*44 Chr.17: 44155294-44155295 on Build GRCh38
472873 c.1717T>A p.Y573N Chr.17: 44155355-44155355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528592557 Chr.17: 44155433-44155433 on Build GRCh38
rs765371857 Chr.17: 44155326-44155326 on Build GRCh38
rs137885797 Chr.17: 44155400-44155400 on Build GRCh38
rs189007 Chr.17: 44155424-44155424 on Build GRCh38
rs372853397 Chr.17: 44155307-44155307 on Build GRCh38
rs192026 Chr.17: 44155359-44155359 on Build GRCh38
rs764466663 Chr.17: 44155413-44155413 on Build GRCh38
rs889442612 Chr.17: 44155418-44155418 on Build GRCh38
rs747432156 Chr.17: 44155373-44155373 on Build GRCh38
rs748985635 Chr.17: 44155292-44155292 on Build GRCh38
rs750283795 Chr.17: 44155442-44155442 on Build GRCh38
rs143469135 Chr.17: 44155401-44155401 on Build GRCh38
rs372663260 Chr.17: 44155362-44155362 on Build GRCh38
rs771289449 Chr.17: 44155380-44155380 on Build GRCh38
rs147545443 Chr.17: 44155363-44155363 on Build GRCh38
rs984078352 Chr.17: 44155423-44155423 on Build GRCh38
rs140080045 Chr.17: 44155364-44155364 on Build GRCh38
rs759467416 Chr.17: 44155311-44155311 on Build GRCh38
rs751393198 Chr.17: 44155428-44155428 on Build GRCh38
rs773544441 Chr.17: 44155372-44155372 on Build GRCh38
rs143332250 Chr.17: 44155357-44155357 on Build GRCh38
rs757243338 Chr.17: 44155339-44155339 on Build GRCh38
rs138929829 Chr.17: 44155327-44155327 on Build GRCh38
rs769486731 Chr.17: 44155258-44155258 on Build GRCh38
rs776760310 Chr.17: 44155310-44155310 on Build GRCh38
rs756169699 Chr.17: 44155432-44155432 on Build GRCh38
rs774942807 Chr.17: 44155271-44155271 on Build GRCh38
rs747153197 Chr.17: 44155428-44155429 on Build GRCh38
rs775676533 Chr.17: 44155407-44155407 on Build GRCh38
rs375554592 Chr.17: 44155383-44155383 on Build GRCh38
rs763241374 Chr.17: 44155411-44155411 on Build GRCh38
rs773801803 Chr.17: 44155427-44155427 on Build GRCh38
rs900951456 Chr.17: 44155405-44155405 on Build GRCh38
rs772381596 Chr.17: 44155294-44155294 on Build GRCh38
rs200296668 Chr.17: 44155328-44155328 on Build GRCh38
rs773599862 Chr.17: 44155296-44155296 on Build GRCh38
rs765189443 Chr.17: 44155406-44155406 on Build GRCh38
rs781341994 Chr.17: 44155356-44155356 on Build GRCh38
rs751446597 Chr.17: 44155334-44155334 on Build GRCh38
rs761142070 Chr.17: 44155300-44155300 on Build GRCh38
rs751338854 Chr.17: 44155308-44155308 on Build GRCh38
rs376118355 Chr.17: 44155422-44155422 on Build GRCh38

View Full Product Details