Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 684763-685004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEAF1
Entrez Gene ID
10522
Gene Name
DEAF1, transcription factor
Gene Aliases
MRD24, NUDR, SPN, ZMYND5
UniGene
Hs.243994
Gene Chromosome Location
Chr.11: 644220-695754 on Build GRCh38
Transcripts
NM_001293634.1 NM_021008.3 XM_011519842.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5831855 c.680C>A p.P227H Chr.11: 684948-684948 on Build GRCh38
5831854 c.681C>T p.P227P Chr.11: 684947-684947 on Build GRCh38
5027336 c.847G>A p.A283T Chr.11: 684921-684921 on Build GRCh38
3810271 c.820C>A p.P274T Chr.11: 684948-684948 on Build GRCh38
3810270 c.821C>T p.P274L Chr.11: 684947-684947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009771106 Chr.11: 684789-684789 on Build GRCh38
rs535060370 Chr.11: 684834-684834 on Build GRCh38
rs140533922 Chr.11: 684910-684910 on Build GRCh38
rs910952535 Chr.11: 684966-684966 on Build GRCh38
rs374363758 Chr.11: 684870-684870 on Build GRCh38
rs774480031 Chr.11: 684977-684977 on Build GRCh38
rs557854316 Chr.11: 684835-684835 on Build GRCh38
rs1034696229 Chr.11: 684855-684855 on Build GRCh38
rs993571544 Chr.11: 684816-684816 on Build GRCh38
rs777450511 Chr.11: 684848-684848 on Build GRCh38
rs547465473 Chr.11: 684849-684849 on Build GRCh38
rs749430059 Chr.11: 684884-684884 on Build GRCh38
rs761093556 Chr.11: 684968-684968 on Build GRCh38
rs868804279 Chr.11: 684959-684959 on Build GRCh38
rs751586843 Chr.11: 684786-684786 on Build GRCh38
rs781619641 Chr.11: 684880-684880 on Build GRCh38
rs533608717 Chr.11: 684896-684896 on Build GRCh38
rs373861624 Chr.11: 684913-684913 on Build GRCh38
rs570273733 Chr.11: 684796-684796 on Build GRCh38
rs925680742 Chr.11: 684984-684984 on Build GRCh38
rs201275407 Chr.11: 684869-684869 on Build GRCh38
rs577899510 Chr.11: 684839-684839 on Build GRCh38
rs979938751 Chr.11: 684812-684812 on Build GRCh38
rs144498322 Chr.11: 684943-684943 on Build GRCh38
rs771073144 Chr.11: 684950-684950 on Build GRCh38
rs370799615 Chr.11: 684885-684885 on Build GRCh38
rs555399097 Chr.11: 684797-684797 on Build GRCh38
rs756331329 Chr.11: 684857-684857 on Build GRCh38
rs73404997 Chr.11: 684798-684798 on Build GRCh38

View Full Product Details