Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97437643-97437870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC1
Entrez Gene ID
51013
Gene Name
exosome component 1
Gene Aliases
CGI-108, CSL4, Csl4p, SKI4, Ski4p, p13
UniGene
Hs.632089
Gene Chromosome Location
Chr.10: 97435909-97446065 on Build GRCh38
Transcripts
NM_001318362.1 NM_001318363.1 NM_001318364.1 NM_001318365.1 NM_001318366.1 NM_016046.4 NR_134586.1 NR_134587.1 XM_011539847.2 XM_017016304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657268 c.370C>T p.P124S Chr.10: 97437726-97437726 on Build GRCh38
7180077 c.349G>C p.E117Q Chr.10: 97437747-97437747 on Build GRCh38
4682057 c.367C>A p.R123S Chr.10: 97437729-97437729 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1029999881 Chr.10: 97437812-97437812 on Build GRCh38
rs958742377 Chr.10: 97437809-97437809 on Build GRCh38
rs376564193 Chr.10: 97437794-97437794 on Build GRCh38
rs1020935235 Chr.10: 97437705-97437705 on Build GRCh38
rs769739512 Chr.10: 97437728-97437728 on Build GRCh38
rs763331706 Chr.10: 97437737-97437738 on Build GRCh38
rs758845405 Chr.10: 97437795-97437795 on Build GRCh38
rs1042341471 Chr.10: 97437702-97437702 on Build GRCh38
rs768860530 Chr.10: 97437707-97437707 on Build GRCh38
rs761705569 Chr.10: 97437710-97437710 on Build GRCh38
rs751220095 Chr.10: 97437750-97437750 on Build GRCh38
rs772598313 Chr.10: 97437766-97437766 on Build GRCh38
rs548480556 Chr.10: 97437815-97437815 on Build GRCh38
rs377660599 Chr.10: 97437788-97437788 on Build GRCh38
rs747168035 Chr.10: 97437701-97437701 on Build GRCh38
rs780444697 Chr.10: 97437680-97437680 on Build GRCh38
rs767075545 Chr.10: 97437753-97437753 on Build GRCh38
rs199945025 Chr.10: 97437676-97437676 on Build GRCh38
rs372178468 Chr.10: 97437804-97437804 on Build GRCh38
rs997285045 Chr.10: 97437808-97437809 on Build GRCh38
rs867804257 Chr.10: 97437716-97437716 on Build GRCh38
rs372810655 Chr.10: 97437734-97437734 on Build GRCh38
rs765780115 Chr.10: 97437672-97437672 on Build GRCh38
rs757740496 Chr.10: 97437755-97437755 on Build GRCh38
rs776846099 Chr.10: 97437709-97437709 on Build GRCh38
rs903760300 Chr.10: 97437729-97437729 on Build GRCh38
rs749830803 Chr.10: 97437670-97437670 on Build GRCh38
rs772985125 Chr.10: 97437738-97437738 on Build GRCh38
rs759188341 Chr.10: 97437751-97437751 on Build GRCh38
rs146531279 Chr.10: 97437740-97437740 on Build GRCh38
rs757765533 Chr.10: 97437675-97437675 on Build GRCh38
rs762830900 Chr.10: 97437739-97437739 on Build GRCh38
rs372971334 Chr.10: 97437754-97437754 on Build GRCh38
rs773549952 Chr.10: 97437814-97437814 on Build GRCh38

View Full Product Details