Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58001457-58001716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USB1
Entrez Gene ID
79650
Gene Name
U6 snRNA biogenesis phosphodiesterase 1
Gene Aliases
C16orf57, HVSL1, Mpn1, PN, hUsb1
UniGene
Hs.408702
Gene Chromosome Location
Chr.16: 57999600-58021623 on Build GRCh38
Transcripts
NM_001195302.1 NM_001204911.1 NM_024598.3 XM_005256144.3 XM_011523328.2 XM_011523329.2 XM_011523330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163558 c.43G>A p.E15K Chr.16: 58001526-58001526 on Build GRCh38
4638716 c.67C>T p.R23W Chr.16: 58001550-58001550 on Build GRCh38
4666979 c.52T>C p.S18P Chr.16: 58001535-58001535 on Build GRCh38
3721028 c.42C>A p.S14S Chr.16: 58001525-58001525 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746264362 Chr.16: 58001537-58001537 on Build GRCh38
rs984082882 Chr.16: 58001627-58001627 on Build GRCh38
rs370555892 Chr.16: 58001476-58001476 on Build GRCh38
rs781769871 Chr.16: 58001626-58001626 on Build GRCh38
rs747958554 Chr.16: 58001544-58001544 on Build GRCh38
rs773091216 Chr.16: 58001479-58001479 on Build GRCh38
rs954060204 Chr.16: 58001501-58001501 on Build GRCh38
rs772792606 Chr.16: 58001557-58001557 on Build GRCh38
rs771539759 Chr.16: 58001551-58001551 on Build GRCh38
rs149411375 Chr.16: 58001573-58001573 on Build GRCh38
rs781151070 Chr.16: 58001496-58001496 on Build GRCh38
rs769698745 Chr.16: 58001623-58001623 on Build GRCh38
rs765469310 Chr.16: 58001498-58001498 on Build GRCh38
rs763219601 Chr.16: 58001607-58001607 on Build GRCh38
rs941753217 Chr.16: 58001481-58001481 on Build GRCh38
rs888896006 Chr.16: 58001480-58001480 on Build GRCh38
rs564572047 Chr.16: 58001506-58001506 on Build GRCh38
rs757242303 Chr.16: 58001624-58001624 on Build GRCh38
rs747805446 Chr.16: 58001550-58001550 on Build GRCh38
rs55807718 Chr.16: 58001643-58001643 on Build GRCh38
rs918644157 Chr.16: 58001571-58001571 on Build GRCh38
rs3743559 Chr.16: 58001525-58001525 on Build GRCh38
rs751660793 Chr.16: 58001616-58001616 on Build GRCh38
rs865993319 Chr.16: 58001568-58001568 on Build GRCh38
rs753636337 Chr.16: 58001489-58001489 on Build GRCh38
rs373657742 Chr.16: 58001503-58001503 on Build GRCh38
rs764990301 Chr.16: 58001584-58001584 on Build GRCh38
rs778756392 Chr.16: 58001544-58001544 on Build GRCh38
rs770232543 Chr.16: 58001539-58001539 on Build GRCh38
rs546288690 Chr.16: 58001486-58001486 on Build GRCh38
rs752987411 Chr.16: 58001596-58001596 on Build GRCh38
rs756585839 Chr.16: 58001633-58001633 on Build GRCh38
rs922885954 Chr.16: 58001684-58001684 on Build GRCh38
rs770934785 Chr.16: 58001563-58001563 on Build GRCh38
rs546677416 Chr.16: 58001662-58001662 on Build GRCh38
rs780553018 Chr.16: 58001636-58001636 on Build GRCh38
rs750942743 Chr.16: 58001628-58001628 on Build GRCh38
rs937581651 Chr.16: 58001691-58001691 on Build GRCh38
rs752060249 Chr.16: 58001514-58001514 on Build GRCh38
rs759290159 Chr.16: 58001496-58001496 on Build GRCh38
rs777899627 Chr.16: 58001513-58001513 on Build GRCh38
rs760648279 Chr.16: 58001484-58001484 on Build GRCh38
rs1046389186 Chr.16: 58001538-58001538 on Build GRCh38
rs759378341 Chr.16: 58001580-58001580 on Build GRCh38
rs966809939 Chr.16: 58001659-58001659 on Build GRCh38
rs976885887 Chr.16: 58001681-58001681 on Build GRCh38
rs1021085102 Chr.16: 58001654-58001654 on Build GRCh38
rs757792250 Chr.16: 58001521-58001521 on Build GRCh38
rs375706004 Chr.16: 58001561-58001561 on Build GRCh38

View Full Product Details