Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11809624-11809883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDFT1
Entrez Gene ID
2222
Gene Name
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases
DGPT, ERG9, SQS, SS
UniGene
Hs.593928
Gene Chromosome Location
Chr.8: 11795573-11839309 on Build GRCh38
Transcripts
NM_001287742.1 NM_001287743.1 NM_001287744.1 NM_001287745.1 NM_001287747.1 NM_001287748.1 NM_001287749.1 NM_001287750.1 NM_001287751.1 NM_001287756.1 NM_004462.4 XM_017013217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5568953 c.265A>G p.S89G Chr.8: 11809734-11809734 on Build GRCh38
127992 c.348G>A p.E116E Chr.8: 11809817-11809817 on Build GRCh38
3763009 c.201C>T p.N67N Chr.8: 11809670-11809670 on Build GRCh38
5160300 c.198-10T>C p.? Chr.8: 11809657-11809657 on Build GRCh38
3896312 c.381+1G>A p.? Chr.8: 11809851-11809851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769598382 Chr.8: 11809795-11809795 on Build GRCh38
rs750494003 Chr.8: 11809845-11809845 on Build GRCh38
rs8417 Chr.8: 11809670-11809670 on Build GRCh38
rs879103973 Chr.8: 11809669-11809669 on Build GRCh38
rs745862193 Chr.8: 11809793-11809793 on Build GRCh38
rs775118817 Chr.8: 11809744-11809744 on Build GRCh38
rs11549152 Chr.8: 11809809-11809809 on Build GRCh38
rs758661215 Chr.8: 11809692-11809692 on Build GRCh38
rs200217031 Chr.8: 11809729-11809729 on Build GRCh38
rs749166948 Chr.8: 11809860-11809860 on Build GRCh38
rs771678941 Chr.8: 11809825-11809825 on Build GRCh38
rs768373931 Chr.8: 11809802-11809802 on Build GRCh38
rs535262343 Chr.8: 11809787-11809787 on Build GRCh38
rs574536561 Chr.8: 11809778-11809778 on Build GRCh38
rs745453575 Chr.8: 11809791-11809791 on Build GRCh38
rs1002449473 Chr.8: 11809808-11809808 on Build GRCh38
rs970626254 Chr.8: 11809788-11809788 on Build GRCh38
rs779973416 Chr.8: 11809850-11809850 on Build GRCh38
rs762645897 Chr.8: 11809748-11809748 on Build GRCh38
rs900240079 Chr.8: 11809723-11809723 on Build GRCh38
rs557080830 Chr.8: 11809829-11809829 on Build GRCh38
rs1005989994 Chr.8: 11809702-11809702 on Build GRCh38
rs61732865 Chr.8: 11809783-11809783 on Build GRCh38
rs774006121 Chr.8: 11809804-11809804 on Build GRCh38
rs775812313 Chr.8: 11809708-11809708 on Build GRCh38
rs754425838 Chr.8: 11809761-11809761 on Build GRCh38
rs777755672 Chr.8: 11809740-11809741 on Build GRCh38
rs111526765 Chr.8: 11809665-11809665 on Build GRCh38
rs367935206 Chr.8: 11809765-11809765 on Build GRCh38
rs201072068 Chr.8: 11809731-11809731 on Build GRCh38
rs773953631 Chr.8: 11809752-11809752 on Build GRCh38
rs753212485 Chr.8: 11809830-11809830 on Build GRCh38
rs371554178 Chr.8: 11809797-11809797 on Build GRCh38
rs149103491 Chr.8: 11809774-11809774 on Build GRCh38
rs368071016 Chr.8: 11809777-11809777 on Build GRCh38
rs900252767 Chr.8: 11809861-11809861 on Build GRCh38
rs759215233 Chr.8: 11809753-11809753 on Build GRCh38
rs751902071 Chr.8: 11809833-11809833 on Build GRCh38
rs1033734483 Chr.8: 11809820-11809820 on Build GRCh38
rs769701350 Chr.8: 11809737-11809737 on Build GRCh38
rs772027071 Chr.8: 11809656-11809657 on Build GRCh38
rs766941596 Chr.8: 11809672-11809672 on Build GRCh38
rs575313773 Chr.8: 11809858-11809858 on Build GRCh38
rs556051417 Chr.8: 11809849-11809849 on Build GRCh38
rs746762581 Chr.8: 11809775-11809776 on Build GRCh38
rs756709654 Chr.8: 11809671-11809671 on Build GRCh38
rs778046889 Chr.8: 11809698-11809698 on Build GRCh38
rs374031686 Chr.8: 11809735-11809735 on Build GRCh38
rs759956045 Chr.8: 11809764-11809764 on Build GRCh38
rs768320911 Chr.8: 11809749-11809749 on Build GRCh38
rs748468392 Chr.8: 11809691-11809691 on Build GRCh38
rs763637687 Chr.8: 11809668-11809668 on Build GRCh38
rs763411837 Chr.8: 11809831-11809831 on Build GRCh38
rs749113831 Chr.8: 11809798-11809798 on Build GRCh38
rs112887229 Chr.8: 11809664-11809664 on Build GRCh38
rs779132325 Chr.8: 11809688-11809688 on Build GRCh38
rs764491287 Chr.8: 11809832-11809832 on Build GRCh38
rs561714716 Chr.8: 11809713-11809713 on Build GRCh38
rs61732866 Chr.8: 11809779-11809779 on Build GRCh38
rs376094577 Chr.8: 11809662-11809662 on Build GRCh38
rs764869946 Chr.8: 11809661-11809661 on Build GRCh38
rs999078339 Chr.8: 11809757-11809757 on Build GRCh38
rs766880676 Chr.8: 11809754-11809754 on Build GRCh38
rs746146450 Chr.8: 11809659-11809659 on Build GRCh38
rs754826172 Chr.8: 11809863-11809863 on Build GRCh38
rs143137518 Chr.8: 11809835-11809835 on Build GRCh38
rs145118363 Chr.8: 11809770-11809770 on Build GRCh38
rs941900174 Chr.8: 11809846-11809846 on Build GRCh38
rs201041296 Chr.8: 11809676-11809676 on Build GRCh38
rs745784138 Chr.8: 11809733-11809733 on Build GRCh38

View Full Product Details