Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126275256-126275503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXRED1
Entrez Gene ID
55572
Gene Name
FAD dependent oxidoreductase domain containing 1
Gene Aliases
FP634, H17
UniGene
Hs.317190
Gene Chromosome Location
Chr.11: 126269040-126278132 on Build GRCh38
Transcripts
NM_017547.3 NR_037647.1 NR_037648.1 XM_006718879.3 XM_006718881.3 XM_011542895.2 XM_011542896.2 XM_017018000.1 XM_017018001.1 XM_017018002.1 XM_017018003.1 XM_017018004.1 XM_017018005.1 XM_017018006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
338952 c.632-1G>C p.? Chr.11: 126275326-126275326 on Build GRCh38
6666579 c.614A>G p.D205G Chr.11: 126275351-126275351 on Build GRCh38
7104967 c.714C>T p.F238F Chr.11: 126275409-126275409 on Build GRCh38
6197998 c.643C>T p.R215* Chr.11: 126275380-126275380 on Build GRCh38
7104968 c.672C>T p.F224F Chr.11: 126275409-126275409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753106152 Chr.11: 126275353-126275353 on Build GRCh38
rs757274475 Chr.11: 126275399-126275399 on Build GRCh38
rs553201412 Chr.11: 126275334-126275334 on Build GRCh38
rs780522584 Chr.11: 126275351-126275352 on Build GRCh38
rs756752516 Chr.11: 126275446-126275446 on Build GRCh38
rs965900698 Chr.11: 126275380-126275380 on Build GRCh38
rs545703077 Chr.11: 126275428-126275428 on Build GRCh38
rs536400690 Chr.11: 126275327-126275327 on Build GRCh38
rs753729232 Chr.11: 126275312-126275312 on Build GRCh38
rs755842141 Chr.11: 126275310-126275310 on Build GRCh38
rs758966293 Chr.11: 126275377-126275377 on Build GRCh38
rs139851379 Chr.11: 126275378-126275378 on Build GRCh38
rs770234474 Chr.11: 126275342-126275342 on Build GRCh38
rs894760353 Chr.11: 126275306-126275306 on Build GRCh38
rs750646991 Chr.11: 126275308-126275309 on Build GRCh38
rs755831680 Chr.11: 126275357-126275357 on Build GRCh38
rs749853207 Chr.11: 126275356-126275356 on Build GRCh38
rs1003912039 Chr.11: 126275312-126275313 on Build GRCh38
rs376645918 Chr.11: 126275309-126275309 on Build GRCh38
rs563695148 Chr.11: 126275462-126275462 on Build GRCh38
rs747044656 Chr.11: 126275328-126275328 on Build GRCh38
rs111517104 Chr.11: 126275467-126275467 on Build GRCh38
rs781203961 Chr.11: 126275337-126275337 on Build GRCh38
rs150562772 Chr.11: 126275370-126275370 on Build GRCh38
rs771536102 Chr.11: 126275465-126275465 on Build GRCh38
rs539113506 Chr.11: 126275387-126275387 on Build GRCh38
rs765078789 Chr.11: 126275400-126275400 on Build GRCh38
rs369054199 Chr.11: 126275463-126275463 on Build GRCh38
rs759285031 Chr.11: 126275301-126275301 on Build GRCh38
rs773703224 Chr.11: 126275298-126275298 on Build GRCh38
rs771320778 Chr.11: 126275364-126275364 on Build GRCh38
rs36109211 Chr.11: 126275386-126275386 on Build GRCh38
rs765873212 Chr.11: 126275296-126275296 on Build GRCh38
rs142594480 Chr.11: 126275338-126275338 on Build GRCh38
rs757107408 Chr.11: 126275315-126275315 on Build GRCh38
rs764036624 Chr.11: 126275311-126275311 on Build GRCh38
rs779224050 Chr.11: 126275471-126275471 on Build GRCh38
rs756374869 Chr.11: 126275459-126275459 on Build GRCh38
rs758853514 Chr.11: 126275326-126275334 on Build GRCh38
rs1035746153 Chr.11: 126275354-126275354 on Build GRCh38
rs754952690 Chr.11: 126275429-126275429 on Build GRCh38
rs775068481 Chr.11: 126275381-126275381 on Build GRCh38
rs781587260 Chr.11: 126275443-126275443 on Build GRCh38
rs267606829 Chr.11: 126275389-126275389 on Build GRCh38
rs752119165 Chr.11: 126275475-126275485 on Build GRCh38

View Full Product Details