Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3015901-3016126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN6
Entrez Gene ID
9074
Gene Name
claudin 6
Gene Aliases
-
UniGene
Hs.533779
Gene Chromosome Location
Chr.16: 3014712-3018187 on Build GRCh38
Transcripts
NM_021195.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6900659 c.14G>A p.G5E Chr.16: 3016008-3016008 on Build GRCh38
4748835 c.88T>C p.W30R Chr.16: 3015934-3015934 on Build GRCh38
471636 c.39G>A p.L13L Chr.16: 3015983-3015983 on Build GRCh38
4672701 c.38T>C p.L13P Chr.16: 3015984-3015984 on Build GRCh38
3817805 c.95T>G p.V32G Chr.16: 3015927-3015927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778629048 Chr.16: 3016048-3016048 on Build GRCh38
rs574214108 Chr.16: 3015960-3015960 on Build GRCh38
rs770429629 Chr.16: 3016043-3016048 on Build GRCh38
rs754199137 Chr.16: 3015944-3015944 on Build GRCh38
rs750988999 Chr.16: 3016010-3016010 on Build GRCh38
rs764277596 Chr.16: 3016003-3016003 on Build GRCh38
rs774150273 Chr.16: 3016052-3016052 on Build GRCh38
rs757564613 Chr.16: 3016008-3016008 on Build GRCh38
rs749095784 Chr.16: 3015975-3015975 on Build GRCh38
rs113281551 Chr.16: 3016025-3016025 on Build GRCh38
rs766294662 Chr.16: 3015922-3015922 on Build GRCh38
rs747727241 Chr.16: 3016024-3016024 on Build GRCh38
rs759021853 Chr.16: 3016016-3016016 on Build GRCh38
rs772914144 Chr.16: 3015967-3015967 on Build GRCh38
rs957586364 Chr.16: 3016001-3016001 on Build GRCh38
rs577456970 Chr.16: 3016020-3016020 on Build GRCh38
rs765594436 Chr.16: 3016009-3016009 on Build GRCh38
rs141047348 Chr.16: 3015937-3015937 on Build GRCh38
rs759627627 Chr.16: 3015929-3015929 on Build GRCh38
rs142284131 Chr.16: 3015989-3015989 on Build GRCh38
rs754029269 Chr.16: 3016005-3016005 on Build GRCh38
rs909605121 Chr.16: 3016094-3016094 on Build GRCh38
rs779297467 Chr.16: 3015946-3015946 on Build GRCh38
rs769609115 Chr.16: 3015965-3015965 on Build GRCh38
rs774161132 Chr.16: 3015982-3015982 on Build GRCh38
rs745569654 Chr.16: 3016064-3016064 on Build GRCh38
rs950483288 Chr.16: 3016086-3016086 on Build GRCh38
rs759431556 Chr.16: 3015988-3015988 on Build GRCh38
rs370100351 Chr.16: 3016029-3016029 on Build GRCh38
rs746316229 Chr.16: 3015947-3015947 on Build GRCh38
rs150258019 Chr.16: 3015943-3015943 on Build GRCh38
rs770664703 Chr.16: 3016051-3016051 on Build GRCh38
rs772041953 Chr.16: 3016070-3016070 on Build GRCh38
rs748905936 Chr.16: 3016039-3016039 on Build GRCh38
rs757624521 Chr.16: 3015945-3015945 on Build GRCh38
rs780798344 Chr.16: 3015956-3015956 on Build GRCh38
rs762777353 Chr.16: 3015921-3015921 on Build GRCh38
rs998939165 Chr.16: 3015969-3015969 on Build GRCh38
rs556923956 Chr.16: 3015996-3015996 on Build GRCh38
rs768311218 Chr.16: 3015954-3015954 on Build GRCh38
rs2269911 Chr.16: 3015923-3015923 on Build GRCh38
rs9940735 Chr.16: 3015980-3015980 on Build GRCh38
rs760716801 Chr.16: 3015997-3015997 on Build GRCh38
rs767520780 Chr.16: 3015932-3015932 on Build GRCh38
rs745642773 Chr.16: 3016066-3016066 on Build GRCh38

View Full Product Details