Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74189509-74189780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4H
Entrez Gene ID
7458
Gene Name
eukaryotic translation initiation factor 4H
Gene Aliases
WBSCR1, WSCR1, eIF-4H
UniGene
Hs.520943
Gene Chromosome Location
Chr.7: 74174376-74197101 on Build GRCh38
Transcripts
NM_022170.1 NM_031992.1 XM_011516556.1 XM_011516557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4389867 c.304G>A p.D102N Chr.7: 74189729-74189729 on Build GRCh38
1091631 c.259G>T p.V87L Chr.7: 74189684-74189684 on Build GRCh38
3367024 c.292G>C p.A98P Chr.7: 74189717-74189717 on Build GRCh38
6472001 c.248-3C>T p.? Chr.7: 74189670-74189670 on Build GRCh38
3640465 c.268G>A p.D90N Chr.7: 74189693-74189693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782820960 Chr.7: 74189728-74189728 on Build GRCh38
rs375960912 Chr.7: 74189648-74189648 on Build GRCh38
rs56820967 Chr.7: 74189581-74189581 on Build GRCh38
rs907205160 Chr.7: 74189597-74189597 on Build GRCh38
rs782520535 Chr.7: 74189746-74189746 on Build GRCh38
rs781887669 Chr.7: 74189734-74189734 on Build GRCh38
rs1045737485 Chr.7: 74189595-74189595 on Build GRCh38
rs782436965 Chr.7: 74189695-74189695 on Build GRCh38
rs575887795 Chr.7: 74189692-74189692 on Build GRCh38
rs782094979 Chr.7: 74189664-74189664 on Build GRCh38
rs370033615 Chr.7: 74189651-74189651 on Build GRCh38
rs191252355 Chr.7: 74189627-74189627 on Build GRCh38
rs782640789 Chr.7: 74189750-74189750 on Build GRCh38
rs940612117 Chr.7: 74189588-74189588 on Build GRCh38
rs781809080 Chr.7: 74189693-74189693 on Build GRCh38
rs782682515 Chr.7: 74189600-74189600 on Build GRCh38
rs782399660 Chr.7: 74189656-74189657 on Build GRCh38
rs984655441 Chr.7: 74189533-74189533 on Build GRCh38
rs781981604 Chr.7: 74189654-74189654 on Build GRCh38
rs1056280287 Chr.7: 74189606-74189606 on Build GRCh38
rs535854917 Chr.7: 74189586-74189586 on Build GRCh38
rs907779856 Chr.7: 74189545-74189545 on Build GRCh38
rs759585992 Chr.7: 74189534-74189534 on Build GRCh38
rs893255755 Chr.7: 74189625-74189625 on Build GRCh38
rs782030365 Chr.7: 74189629-74189629 on Build GRCh38

View Full Product Details