Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1448522-1448755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN7
Entrez Gene ID
1186
Gene Name
chloride voltage-gated channel 7
Gene Aliases
CLC-7, CLC7, OPTA2, OPTB4, PPP1R63
UniGene
Hs.459649
Gene Chromosome Location
Chr.16: 1444933-1475084 on Build GRCh38
Transcripts
NM_001114331.2 NM_001287.5 XM_011522354.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629905 c.1843C>T p.H615Y Chr.16: 1448721-1448721 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs17135338 Chr.16: 1448571-1448571 on Build GRCh38
rs79477995 Chr.16: 1448596-1448596 on Build GRCh38
rs570443272 Chr.16: 1448609-1448609 on Build GRCh38
rs990862714 Chr.16: 1448569-1448569 on Build GRCh38
rs772244670 Chr.16: 1448687-1448687 on Build GRCh38
rs1000353389 Chr.16: 1448724-1448724 on Build GRCh38
rs564558076 Chr.16: 1448547-1448547 on Build GRCh38
rs78002466 Chr.16: 1448578-1448578 on Build GRCh38
rs370045081 Chr.16: 1448719-1448719 on Build GRCh38
rs375830369 Chr.16: 1448701-1448701 on Build GRCh38
rs778590639 Chr.16: 1448646-1448646 on Build GRCh38
rs371416385 Chr.16: 1448670-1448670 on Build GRCh38
rs749814550 Chr.16: 1448671-1448671 on Build GRCh38
rs754928380 Chr.16: 1448638-1448638 on Build GRCh38
rs149403063 Chr.16: 1448734-1448734 on Build GRCh38
rs150643398 Chr.16: 1448584-1448584 on Build GRCh38
rs942884737 Chr.16: 1448608-1448608 on Build GRCh38
rs540469959 Chr.16: 1448568-1448568 on Build GRCh38
rs759560642 Chr.16: 1448730-1448730 on Build GRCh38
rs561369777 Chr.16: 1448591-1448591 on Build GRCh38
rs769089903 Chr.16: 1448673-1448673 on Build GRCh38
rs988502611 Chr.16: 1448582-1448582 on Build GRCh38
rs773464510 Chr.16: 1448700-1448700 on Build GRCh38
rs746266169 Chr.16: 1448658-1448658 on Build GRCh38
rs369389740 Chr.16: 1448660-1448660 on Build GRCh38
rs139826207 Chr.16: 1448707-1448707 on Build GRCh38
rs780354249 Chr.16: 1448665-1448665 on Build GRCh38
rs187719297 Chr.16: 1448602-1448602 on Build GRCh38
rs372691390 Chr.16: 1448674-1448674 on Build GRCh38
rs543067738 Chr.16: 1448561-1448561 on Build GRCh38
rs753798178 Chr.16: 1448630-1448630 on Build GRCh38
rs1037040048 Chr.16: 1448663-1448663 on Build GRCh38
rs201396352 Chr.16: 1448676-1448676 on Build GRCh38
rs893520241 Chr.16: 1448627-1448627 on Build GRCh38
rs907822473 Chr.16: 1448629-1448629 on Build GRCh38
rs777468989 Chr.16: 1448654-1448654 on Build GRCh38
rs533199941 Chr.16: 1448617-1448617 on Build GRCh38
rs757991875 Chr.16: 1448647-1448647 on Build GRCh38
rs972719884 Chr.16: 1448583-1448583 on Build GRCh38

View Full Product Details