Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184249532-184249781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECE2
Entrez Gene ID
9718
Gene Name
endothelin converting enzyme 2
Gene Aliases
-
UniGene
Hs.146161
Gene Chromosome Location
Chr.3: 184249657-184293031 on Build GRCh38
Transcripts
NM_014693.3 NM_032331.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3714501 c.32C>T p.P11L Chr.3: 184249726-184249726 on Build GRCh38
4157469 c.47G>T p.R16L Chr.3: 184249741-184249741 on Build GRCh38
5179662 c.18A>C p.A6A Chr.3: 184249712-184249712 on Build GRCh38
6651266 c.62G>A p.R21H Chr.3: 184249756-184249756 on Build GRCh38
5179664 c.19G>A p.G7S Chr.3: 184249713-184249713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768780282 Chr.3: 184249695-184249695 on Build GRCh38
rs761335842 Chr.3: 184249763-184249763 on Build GRCh38
rs763705935 Chr.3: 184249702-184249702 on Build GRCh38
rs200184637 Chr.3: 184249732-184249732 on Build GRCh38
rs774743971 Chr.3: 184249742-184249742 on Build GRCh38
rs886585179 Chr.3: 184249611-184249611 on Build GRCh38
rs776078211 Chr.3: 184249757-184249757 on Build GRCh38
rs113457645 Chr.3: 184249714-184249714 on Build GRCh38
rs559606291 Chr.3: 184249678-184249678 on Build GRCh38
rs71632014 Chr.3: 184249710-184249710 on Build GRCh38
rs756709771 Chr.3: 184249683-184249683 on Build GRCh38
rs779918442 Chr.3: 184249719-184249719 on Build GRCh38
rs942571836 Chr.3: 184249617-184249617 on Build GRCh38
rs766759873 Chr.3: 184249690-184249690 on Build GRCh38
rs954792202 Chr.3: 184249564-184249564 on Build GRCh38
rs1009713077 Chr.3: 184249751-184249751 on Build GRCh38
rs558939975 Chr.3: 184249641-184249641 on Build GRCh38
rs200563134 Chr.3: 184249729-184249729 on Build GRCh38
rs111287000 Chr.3: 184249681-184249681 on Build GRCh38
rs774634317 Chr.3: 184249691-184249691 on Build GRCh38
rs111425351 Chr.3: 184249666-184249666 on Build GRCh38
rs898187649 Chr.3: 184249596-184249596 on Build GRCh38
rs200612790 Chr.3: 184249669-184249669 on Build GRCh38
rs772653030 Chr.3: 184249578-184249578 on Build GRCh38
rs755633384 Chr.3: 184249701-184249701 on Build GRCh38
rs374014207 Chr.3: 184249698-184249698 on Build GRCh38
rs773230528 Chr.3: 184249725-184249725 on Build GRCh38
rs987846934 Chr.3: 184249583-184249583 on Build GRCh38
rs769797764 Chr.3: 184249722-184249722 on Build GRCh38
rs773364920 Chr.3: 184249673-184249673 on Build GRCh38
rs768012597 Chr.3: 184249748-184249748 on Build GRCh38
rs753528254 Chr.3: 184249706-184249706 on Build GRCh38
rs546171403 Chr.3: 184249592-184249592 on Build GRCh38
rs959391163 Chr.3: 184249642-184249642 on Build GRCh38
rs76486175 Chr.3: 184249707-184249707 on Build GRCh38
rs748186983 Chr.3: 184249667-184249667 on Build GRCh38
rs995235274 Chr.3: 184249601-184249601 on Build GRCh38
rs759775871 Chr.3: 184249747-184249747 on Build GRCh38
rs919752675 Chr.3: 184249623-184249623 on Build GRCh38
rs1046899225 Chr.3: 184249648-184249648 on Build GRCh38
rs1021075573 Chr.3: 184249553-184249553 on Build GRCh38
rs931175452 Chr.3: 184249624-184249624 on Build GRCh38
rs771105994 Chr.3: 184249740-184249740 on Build GRCh38
rs975393634 Chr.3: 184249618-184249618 on Build GRCh38
rs2233461 Chr.3: 184249712-184249712 on Build GRCh38
rs968454889 Chr.3: 184249689-184249689 on Build GRCh38
rs780567276 Chr.3: 184249605-184249605 on Build GRCh38
rs1028986550 Chr.3: 184249682-184249682 on Build GRCh38
rs143159008 Chr.3: 184249720-184249720 on Build GRCh38
rs1044730503 Chr.3: 184249586-184249586 on Build GRCh38

View Full Product Details